Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18813622-18813895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf78
Entrez Gene ID
100128496
Gene Name
chromosome 20 open reading frame 78
Gene Aliases
dJ1068E13.1
UniGene
Hs.664484
Gene Chromosome Location
Chr.20: 18809351-18830203 on Build GRCh38
Transcripts
NM_001242671.2 NM_001302818.1 NM_178483.2

Gene Symbol
SCP2D1
Entrez Gene ID
140856
Gene Name
SCP2 sterol binding domain containing 1
Gene Aliases
C20orf79, HSD22, dJ1068E13.2
UniGene
Hs.112794
Gene Chromosome Location
Chr.20: 18813726-18814392 on Build GRCh38
Transcripts
NM_001242671.2 NM_001302818.1 NM_178483.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4921673 c.58G>T p.G20C Chr.20: 18813873-18813873 on Build GRCh38
4901320 c.66C>T p.F22F Chr.20: 18813881-18813881 on Build GRCh38
4800414 c.67G>T p.E23* Chr.20: 18813882-18813882 on Build GRCh38
5578174 c.67G>A p.E23K Chr.20: 18813882-18813882 on Build GRCh38
5926085 c.47G>A p.G16E Chr.20: 18813862-18813862 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776432081 Chr.20: 18813870-18813870 on Build GRCh38
rs756453842 Chr.20: 18813792-18813792 on Build GRCh38
rs201406631 Chr.20: 18813818-18813818 on Build GRCh38
rs531449512 Chr.20: 18813685-18813685 on Build GRCh38
rs547630005 Chr.20: 18813784-18813784 on Build GRCh38
rs780421406 Chr.20: 18813803-18813803 on Build GRCh38
rs879329259 Chr.20: 18813733-18813733 on Build GRCh38
rs750846012 Chr.20: 18813786-18813786 on Build GRCh38
rs746925152 Chr.20: 18813841-18813841 on Build GRCh38
rs1020268431 Chr.20: 18813720-18813720 on Build GRCh38
rs988791011 Chr.20: 18813717-18813717 on Build GRCh38
rs879075876 Chr.20: 18813703-18813703 on Build GRCh38
rs781006204 Chr.20: 18813813-18813813 on Build GRCh38
rs930923636 Chr.20: 18813660-18813660 on Build GRCh38
rs568111212 Chr.20: 18813750-18813750 on Build GRCh38
rs527842256 Chr.20: 18813651-18813651 on Build GRCh38
rs757831197 Chr.20: 18813770-18813770 on Build GRCh38
rs748896831 Chr.20: 18813813-18813813 on Build GRCh38
rs996710311 Chr.20: 18813699-18813699 on Build GRCh38
rs925190696 Chr.20: 18813769-18813769 on Build GRCh38
rs368681702 Chr.20: 18813833-18813833 on Build GRCh38
rs1016915375 Chr.20: 18813687-18813687 on Build GRCh38
rs59741823 Chr.20: 18813656-18813656 on Build GRCh38
rs760825661 Chr.20: 18813659-18813659 on Build GRCh38
rs954764108 Chr.20: 18813712-18813712 on Build GRCh38
rs773306131 Chr.20: 18813825-18813825 on Build GRCh38
rs764475930 Chr.20: 18813691-18813691 on Build GRCh38
rs917683341 Chr.20: 18813797-18813797 on Build GRCh38
rs772186682 Chr.20: 18813824-18813824 on Build GRCh38
rs1013263735 Chr.20: 18813650-18813650 on Build GRCh38
rs968680039 Chr.20: 18813726-18813726 on Build GRCh38
rs763146845 Chr.20: 18813871-18813871 on Build GRCh38
rs749940712 Chr.20: 18813844-18813844 on Build GRCh38
rs757786265 Chr.20: 18813785-18813785 on Build GRCh38
rs41308631 Chr.20: 18813819-18813819 on Build GRCh38
rs138003057 Chr.20: 18813713-18813713 on Build GRCh38
rs564180497 Chr.20: 18813674-18813674 on Build GRCh38
rs747654523 Chr.20: 18813823-18813823 on Build GRCh38
rs931986466 Chr.20: 18813767-18813767 on Build GRCh38
rs979408731 Chr.20: 18813751-18813751 on Build GRCh38
rs903776476 Chr.20: 18813817-18813817 on Build GRCh38
rs41308066 Chr.20: 18813773-18813773 on Build GRCh38

View Full Product Details