Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53912931-53913156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG7
Entrez Gene ID
59284
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 7
Gene Aliases
-
UniGene
Hs.631597
Gene Chromosome Location
Chr.19: 53909425-53943944 on Build GRCh38
Transcripts
NM_031896.4 XM_005259124.3 XM_017027093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3389330 c.128C>T p.P43L Chr.19: 53912959-53912959 on Build GRCh38
1247205 c.115G>A p.G39S Chr.19: 53912946-53912946 on Build GRCh38
6525347 c.136C>A p.Q46K Chr.19: 53912967-53912967 on Build GRCh38
4081143 c.107T>C p.M36T Chr.19: 53912938-53912938 on Build GRCh38
309819 c.189C>G p.F63L Chr.19: 53913020-53913020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758861033 Chr.19: 53913040-53913041 on Build GRCh38
rs748218872 Chr.19: 53913048-53913048 on Build GRCh38
rs778081875 Chr.19: 53913058-53913058 on Build GRCh38
rs763476586 Chr.19: 53912994-53912994 on Build GRCh38
rs932376482 Chr.19: 53913031-53913031 on Build GRCh38
rs532242896 Chr.19: 53913012-53913012 on Build GRCh38
rs1000309496 Chr.19: 53913129-53913129 on Build GRCh38
rs1001926777 Chr.19: 53913030-53913030 on Build GRCh38
rs909578331 Chr.19: 53913055-53913056 on Build GRCh38
rs747290529 Chr.19: 53913066-53913066 on Build GRCh38
rs779891982 Chr.19: 53913041-53913041 on Build GRCh38
rs546530087 Chr.19: 53912955-53912955 on Build GRCh38
rs778738834 Chr.19: 53913047-53913047 on Build GRCh38
rs939679965 Chr.19: 53913055-53913055 on Build GRCh38
rs189778120 Chr.19: 53912976-53912976 on Build GRCh38
rs370400432 Chr.19: 53912975-53912975 on Build GRCh38
rs371890578 Chr.19: 53913042-53913042 on Build GRCh38
rs1014089959 Chr.19: 53913098-53913098 on Build GRCh38
rs367634340 Chr.19: 53912999-53912999 on Build GRCh38
rs376239971 Chr.19: 53912960-53912960 on Build GRCh38
rs773567206 Chr.19: 53913076-53913076 on Build GRCh38
rs569022701 Chr.19: 53913088-53913088 on Build GRCh38
rs61664013 Chr.19: 53913086-53913086 on Build GRCh38
rs769846106 Chr.19: 53912959-53912959 on Build GRCh38
rs763263137 Chr.19: 53913110-53913110 on Build GRCh38
rs73937618 Chr.19: 53913026-53913026 on Build GRCh38
rs1024570557 Chr.19: 53913021-53913021 on Build GRCh38
rs896056577 Chr.19: 53912997-53912997 on Build GRCh38
rs374846922 Chr.19: 53912996-53912996 on Build GRCh38
rs28580646 Chr.19: 53913102-53913102 on Build GRCh38
rs758555876 Chr.19: 53913054-53913054 on Build GRCh38
rs755892213 Chr.19: 53913035-53913035 on Build GRCh38
rs895690087 Chr.19: 53913094-53913094 on Build GRCh38
rs376073772 Chr.19: 53913043-53913043 on Build GRCh38
rs776346962 Chr.19: 53913052-53913052 on Build GRCh38
rs905959645 Chr.19: 53913111-53913111 on Build GRCh38
rs745649137 Chr.19: 53912986-53912986 on Build GRCh38
rs767093695 Chr.19: 53913002-53913002 on Build GRCh38
rs987390052 Chr.19: 53913089-53913089 on Build GRCh38

View Full Product Details