Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80161846-80162117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA11
Entrez Gene ID
311
Gene Name
annexin A11
Gene Aliases
ANX11, CAP50
UniGene
Hs.530291
Gene Chromosome Location
Chr.10: 80153953-80205677 on Build GRCh38
Transcripts
NM_001157.2 NM_001278407.1 NM_001278408.1 NM_001278409.1 NM_145868.1 NM_145869.1 XM_005269741.4 XM_005269742.1 XM_006717813.1 XM_006717814.3 XM_011539735.1 XM_011539736.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6032528 c.1126G>A p.E376K Chr.10: 80161989-80161989 on Build GRCh38
6841551 c.1111C>T p.R371C Chr.10: 80162004-80162004 on Build GRCh38
7233904 c.1105G>A p.E369K Chr.10: 80162010-80162010 on Build GRCh38
6841552 c.9C>T p.N3N Chr.10: 80162004-80162004 on Build GRCh38
5518989 c.1097C>T p.A366V Chr.10: 80162018-80162018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747033001 Chr.10: 80162072-80162072 on Build GRCh38
rs1049332449 Chr.10: 80162070-80162070 on Build GRCh38
rs749857234 Chr.10: 80161913-80161913 on Build GRCh38
rs147829571 Chr.10: 80161989-80161989 on Build GRCh38
rs1046909585 Chr.10: 80161876-80161876 on Build GRCh38
rs34414015 Chr.10: 80162010-80162010 on Build GRCh38
rs760284084 Chr.10: 80161938-80161941 on Build GRCh38
rs756835280 Chr.10: 80161904-80161904 on Build GRCh38
rs573836158 Chr.10: 80162089-80162089 on Build GRCh38
rs746081543 Chr.10: 80162011-80162011 on Build GRCh38
rs749966764 Chr.10: 80161976-80161976 on Build GRCh38
rs375046856 Chr.10: 80161886-80161886 on Build GRCh38
rs76666369 Chr.10: 80162036-80162036 on Build GRCh38
rs200674282 Chr.10: 80162031-80162031 on Build GRCh38
rs887608267 Chr.10: 80162075-80162075 on Build GRCh38
rs149698938 Chr.10: 80161866-80161866 on Build GRCh38
rs567531642 Chr.10: 80162077-80162078 on Build GRCh38
rs780307887 Chr.10: 80162069-80162069 on Build GRCh38
rs772333853 Chr.10: 80162013-80162013 on Build GRCh38
rs757839642 Chr.10: 80161979-80161979 on Build GRCh38
rs757513641 Chr.10: 80162004-80162004 on Build GRCh38
rs755243184 Chr.10: 80161996-80161996 on Build GRCh38
rs762584554 Chr.10: 80162038-80162038 on Build GRCh38
rs904291188 Chr.10: 80161956-80161956 on Build GRCh38
rs764839772 Chr.10: 80161908-80161908 on Build GRCh38
rs780256561 Chr.10: 80162014-80162014 on Build GRCh38
rs560541777 Chr.10: 80161874-80161874 on Build GRCh38
rs779912437 Chr.10: 80161917-80161917 on Build GRCh38
rs770460417 Chr.10: 80161989-80161990 on Build GRCh38
rs1055965178 Chr.10: 80162023-80162023 on Build GRCh38
rs758231628 Chr.10: 80161955-80161955 on Build GRCh38
rs908414775 Chr.10: 80161895-80161895 on Build GRCh38
rs762030128 Chr.10: 80162021-80162021 on Build GRCh38
rs758358581 Chr.10: 80161916-80161916 on Build GRCh38
rs776413311 Chr.10: 80161990-80161991 on Build GRCh38
rs372196644 Chr.10: 80161952-80161952 on Build GRCh38
rs769495145 Chr.10: 80162029-80162029 on Build GRCh38
rs764546727 Chr.10: 80161891-80161891 on Build GRCh38
rs763214442 Chr.10: 80161992-80161993 on Build GRCh38
rs781488814 Chr.10: 80162076-80162076 on Build GRCh38
rs113463944 Chr.10: 80161886-80161887 on Build GRCh38
rs746813362 Chr.10: 80161929-80161929 on Build GRCh38
rs146222704 Chr.10: 80161959-80161959 on Build GRCh38
rs539605721 Chr.10: 80161953-80161953 on Build GRCh38
rs765925468 Chr.10: 80162044-80162044 on Build GRCh38
rs1017512651 Chr.10: 80162090-80162090 on Build GRCh38
rs755927816 Chr.10: 80162051-80162051 on Build GRCh38
rs780837868 Chr.10: 80161936-80161936 on Build GRCh38
rs753582528 Chr.10: 80161897-80161897 on Build GRCh38
rs368776217 Chr.10: 80162047-80162047 on Build GRCh38
rs370575501 Chr.10: 80162018-80162018 on Build GRCh38
rs555993142 Chr.10: 80161997-80161997 on Build GRCh38
rs938586921 Chr.10: 80161906-80161906 on Build GRCh38
rs964708003 Chr.10: 80162020-80162020 on Build GRCh38
rs747556311 Chr.10: 80162015-80162015 on Build GRCh38
rs2304410 Chr.10: 80162054-80162054 on Build GRCh38
rs143165716 Chr.10: 80161972-80161972 on Build GRCh38
rs781081340 Chr.10: 80161998-80161998 on Build GRCh38
rs751807530 Chr.10: 80161990-80161990 on Build GRCh38
rs894185702 Chr.10: 80161947-80161947 on Build GRCh38
rs749573800 Chr.10: 80161946-80161946 on Build GRCh38
rs141456938 Chr.10: 80162017-80162017 on Build GRCh38
rs45557433 Chr.10: 80162046-80162046 on Build GRCh38
rs563638935 Chr.10: 80162087-80162087 on Build GRCh38
rs367988681 Chr.10: 80161888-80161888 on Build GRCh38
rs993364547 Chr.10: 80162059-80162059 on Build GRCh38
rs769223181 Chr.10: 80162074-80162074 on Build GRCh38
rs199963158 Chr.10: 80161966-80161966 on Build GRCh38
rs577767386 Chr.10: 80162052-80162052 on Build GRCh38
rs776010573 Chr.10: 80161884-80161884 on Build GRCh38
rs766027057 Chr.10: 80161975-80161975 on Build GRCh38
rs367579997 Chr.10: 80162068-80162068 on Build GRCh38
rs376869591 Chr.10: 80161958-80161958 on Build GRCh38
rs139186583 Chr.10: 80162003-80162003 on Build GRCh38
rs547460380 Chr.10: 80161933-80161933 on Build GRCh38

View Full Product Details