Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232024154-232024415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIS3L2
Entrez Gene ID
129563
Gene Name
DIS3 like 3'-5' exoribonuclease 2
Gene Aliases
FAM6A, PRLMNS, hDIS3L2
UniGene
Hs.732236
Gene Chromosome Location
Chr.2: 231961583-232343968 on Build GRCh38
Transcripts
NM_001257281.1 NM_001257282.1 NM_152383.4 NR_046476.1 NR_046477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6644345 c.264G>A p.P88P Chr.2: 232024330-232024330 on Build GRCh38
4931360 c.231A>T p.P77P Chr.2: 232024297-232024297 on Build GRCh38
6644344 c.264G>A p.P88P Chr.2: 232024330-232024330 on Build GRCh38
5938855 c.211-1G>A p.? Chr.2: 232024276-232024276 on Build GRCh38
6644341 c.257C>A p.P86H Chr.2: 232024323-232024323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1036515873 Chr.2: 232024351-232024351 on Build GRCh38
rs377020708 Chr.2: 232024327-232024327 on Build GRCh38
rs113426398 Chr.2: 232024314-232024314 on Build GRCh38
rs908679435 Chr.2: 232024215-232024215 on Build GRCh38
rs767260895 Chr.2: 232024261-232024262 on Build GRCh38
rs769166298 Chr.2: 232024348-232024348 on Build GRCh38
rs373728819 Chr.2: 232024350-232024350 on Build GRCh38
rs753272980 Chr.2: 232024366-232024366 on Build GRCh38
rs771333993 Chr.2: 232024359-232024359 on Build GRCh38
rs867217314 Chr.2: 232024284-232024284 on Build GRCh38
rs914019616 Chr.2: 232024221-232024221 on Build GRCh38
rs377110632 Chr.2: 232024361-232024361 on Build GRCh38
rs745443608 Chr.2: 232024339-232024339 on Build GRCh38
rs925286669 Chr.2: 232024181-232024181 on Build GRCh38
rs544679412 Chr.2: 232024214-232024214 on Build GRCh38
rs751230116 Chr.2: 232024323-232024323 on Build GRCh38
rs555061413 Chr.2: 232024352-232024352 on Build GRCh38
rs774221188 Chr.2: 232024353-232024353 on Build GRCh38
rs773590205 Chr.2: 232024230-232024230 on Build GRCh38
rs753108965 Chr.2: 232024282-232024282 on Build GRCh38
rs887424420 Chr.2: 232024330-232024330 on Build GRCh38
rs938276359 Chr.2: 232024325-232024325 on Build GRCh38
rs373997654 Chr.2: 232024218-232024218 on Build GRCh38
rs936747278 Chr.2: 232024202-232024202 on Build GRCh38
rs759518693 Chr.2: 232024274-232024274 on Build GRCh38
rs369708165 Chr.2: 232024238-232024238 on Build GRCh38
rs761693564 Chr.2: 232024357-232024357 on Build GRCh38
rs952399615 Chr.2: 232024192-232024192 on Build GRCh38
rs200460227 Chr.2: 232024295-232024295 on Build GRCh38
rs373861286 Chr.2: 232024303-232024303 on Build GRCh38
rs754036208 Chr.2: 232024261-232024261 on Build GRCh38
rs766569992 Chr.2: 232024257-232024257 on Build GRCh38
rs765818092 Chr.2: 232024280-232024280 on Build GRCh38
rs202059499 Chr.2: 232024329-232024329 on Build GRCh38
rs893912059 Chr.2: 232024287-232024287 on Build GRCh38
rs765195518 Chr.2: 232024364-232024364 on Build GRCh38
rs752596889 Chr.2: 232024268-232024268 on Build GRCh38
rs376617215 Chr.2: 232024254-232024254 on Build GRCh38
rs377642822 Chr.2: 232024229-232024229 on Build GRCh38
rs984304436 Chr.2: 232024206-232024206 on Build GRCh38

View Full Product Details