Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210022734-210022995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANSL1L
Entrez Gene ID
151050
Gene Name
KAT8 regulatory NSL complex subunit 1 like
Gene Aliases
C2orf67, MSL1v2
UniGene
Hs.282260
Gene Chromosome Location
Chr.2: 210021421-210171370 on Build GRCh38
Transcripts
NM_001307976.1 NM_152519.3 XM_005246328.4 XM_005246329.4 XM_005246330.4 XM_005246332.4 XM_011510706.2 XM_011510707.2 XM_011510710.2 XM_017003430.1 XM_017003431.1 XM_006712677.3

Gene Symbol
RPE
Entrez Gene ID
6120
Gene Name
ribulose-5-phosphate-3-epimerase
Gene Aliases
RPE2-1
UniGene
Hs.282260
Gene Chromosome Location
Chr.2: 210002565-210029156 on Build GRCh38
Transcripts
NM_001307976.1 NM_152519.3 XM_005246328.4 XM_005246329.4 XM_005246330.4 XM_005246332.4 XM_011510706.2 XM_011510707.2 XM_011510710.2 XM_017003430.1 XM_017003431.1 XM_006712677.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6690080 c.2947G>A p.V983I Chr.2: 210022966-210022966 on Build GRCh38
1163721 c.2957delA p.N986fs*>2 Chr.2: 210022956-210022956 on Build GRCh38
220146 c.2959A>G p.R987G Chr.2: 210022954-210022954 on Build GRCh38
4090752 c.2918A>T p.Y973F Chr.2: 210022995-210022995 on Build GRCh38
5474389 c.2961G>A p.R987R Chr.2: 210022952-210022952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555517490 Chr.2: 210022944-210022944 on Build GRCh38
rs951871964 Chr.2: 210022795-210022795 on Build GRCh38
rs374275064 Chr.2: 210022906-210022906 on Build GRCh38
rs1052534306 Chr.2: 210022878-210022878 on Build GRCh38
rs576309982 Chr.2: 210022917-210022917 on Build GRCh38
rs867386313 Chr.2: 210022929-210022929 on Build GRCh38
rs761934507 Chr.2: 210022913-210022913 on Build GRCh38
rs772969356 Chr.2: 210022905-210022905 on Build GRCh38
rs989739630 Chr.2: 210022833-210022833 on Build GRCh38
rs747758543 Chr.2: 210022899-210022899 on Build GRCh38
rs980880357 Chr.2: 210022761-210022761 on Build GRCh38
rs556627515 Chr.2: 210022889-210022889 on Build GRCh38
rs539511619 Chr.2: 210022836-210022836 on Build GRCh38
rs376580380 Chr.2: 210022824-210022824 on Build GRCh38
rs892760815 Chr.2: 210022879-210022879 on Build GRCh38
rs903574590 Chr.2: 210022854-210022854 on Build GRCh38
rs867173633 Chr.2: 210022859-210022859 on Build GRCh38
rs565963819 Chr.2: 210022808-210022808 on Build GRCh38
rs765902812 Chr.2: 210022939-210022939 on Build GRCh38
rs191864182 Chr.2: 210022961-210022961 on Build GRCh38
rs757475166 Chr.2: 210022966-210022966 on Build GRCh38
rs377217595 Chr.2: 210022955-210022955 on Build GRCh38
rs769695051 Chr.2: 210022955-210022956 on Build GRCh38
rs982091404 Chr.2: 210022851-210022851 on Build GRCh38
rs747849946 Chr.2: 210022956-210022956 on Build GRCh38
rs969324722 Chr.2: 210022838-210022838 on Build GRCh38
rs187051510 Chr.2: 210022811-210022811 on Build GRCh38
rs919982359 Chr.2: 210022791-210022791 on Build GRCh38
rs150622688 Chr.2: 210022938-210022938 on Build GRCh38
rs10203742 Chr.2: 210022819-210022819 on Build GRCh38
rs771385451 Chr.2: 210022907-210022907 on Build GRCh38
rs984606800 Chr.2: 210022801-210022802 on Build GRCh38
rs927982796 Chr.2: 210022873-210022873 on Build GRCh38

View Full Product Details