Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19356558-19356793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZB
Entrez Gene ID
832
Gene Name
capping actin protein of muscle Z-line beta subunit
Gene Aliases
CAPB, CAPPB, CAPZ
UniGene
Hs.432760
Gene Chromosome Location
Chr.1: 19338773-19485641 on Build GRCh38
Transcripts
NM_001206540.2 NM_001206541.2 NM_001282162.1 NM_004930.4 XM_006710938.3 XM_011542228.2 XM_011542229.2 XM_011542230.2 XM_017002428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6421830 c.665A>G p.Q222R Chr.1: 19356636-19356636 on Build GRCh38
6042174 c.511T>G p.S171A Chr.1: 19356712-19356712 on Build GRCh38
7278115 c.575G>A p.R192H Chr.1: 19356735-19356735 on Build GRCh38
6378467 c.631G>T p.G211C Chr.1: 19356670-19356670 on Build GRCh38
6042173 c.511T>G p.S171A Chr.1: 19356712-19356712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374636744 Chr.1: 19356619-19356619 on Build GRCh38
rs765706838 Chr.1: 19356676-19356676 on Build GRCh38
rs370957079 Chr.1: 19356708-19356708 on Build GRCh38
rs762321251 Chr.1: 19356667-19356667 on Build GRCh38
rs376741149 Chr.1: 19356631-19356631 on Build GRCh38
rs775456434 Chr.1: 19356622-19356622 on Build GRCh38
rs778714039 Chr.1: 19356739-19356739 on Build GRCh38
rs773641339 Chr.1: 19356707-19356707 on Build GRCh38
rs563349284 Chr.1: 19356774-19356774 on Build GRCh38
rs532731972 Chr.1: 19356732-19356732 on Build GRCh38
rs143868401 Chr.1: 19356734-19356734 on Build GRCh38
rs752190963 Chr.1: 19356725-19356725 on Build GRCh38
rs1012732497 Chr.1: 19356679-19356679 on Build GRCh38
rs868786703 Chr.1: 19356768-19356768 on Build GRCh38
rs935456912 Chr.1: 19356592-19356592 on Build GRCh38
rs377121153 Chr.1: 19356736-19356736 on Build GRCh38
rs766890226 Chr.1: 19356722-19356722 on Build GRCh38
rs572934556 Chr.1: 19356594-19356594 on Build GRCh38
rs747087349 Chr.1: 19356623-19356623 on Build GRCh38
rs776907890 Chr.1: 19356653-19356653 on Build GRCh38
rs373170769 Chr.1: 19356731-19356731 on Build GRCh38
rs745616037 Chr.1: 19356747-19356747 on Build GRCh38
rs745810413 Chr.1: 19356607-19356607 on Build GRCh38

View Full Product Details