Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 145635280-145635521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPM2A
Entrez Gene ID
7957
Gene Name
epilepsy, progressive myoclonus type 2A, Lafora disease (laforin)
Gene Aliases
EPM2, MELF
UniGene
Hs.486696
Gene Chromosome Location
Chr.6: 145500744-145736018 on Build GRCh38
Transcripts
NM_001018041.1 NM_005670.3 XM_006715564.3 XM_011536113.2 XM_011536116.1 XM_017011300.1 XM_017011301.1 XM_017011302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4681390 c.591T>C p.D197D Chr.6: 145635372-145635372 on Build GRCh38
5404518 c.331C>T p.P111S Chr.6: 145635331-145635331 on Build GRCh38
4762369 c.544C>T p.H182Y Chr.6: 145635419-145635419 on Build GRCh38
4948812 c.332C>A p.P111H Chr.6: 145635330-145635330 on Build GRCh38
7154171 c.348G>C p.W116C Chr.6: 145635374-145635374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141919651 Chr.6: 145635475-145635475 on Build GRCh38
rs749100678 Chr.6: 145635491-145635491 on Build GRCh38
rs777767978 Chr.6: 145635476-145635476 on Build GRCh38
rs758599535 Chr.6: 145635306-145635306 on Build GRCh38
rs375544596 Chr.6: 145635382-145635382 on Build GRCh38
rs571938170 Chr.6: 145635343-145635343 on Build GRCh38
rs771562490 Chr.6: 145635371-145635371 on Build GRCh38
rs186710183 Chr.6: 145635344-145635344 on Build GRCh38
rs983510084 Chr.6: 145635398-145635398 on Build GRCh38
rs780241344 Chr.6: 145635318-145635318 on Build GRCh38
rs147645370 Chr.6: 145635467-145635467 on Build GRCh38
rs765641184 Chr.6: 145635396-145635396 on Build GRCh38
rs199856913 Chr.6: 145635473-145635473 on Build GRCh38
rs950789713 Chr.6: 145635393-145635393 on Build GRCh38
rs909269002 Chr.6: 145635413-145635413 on Build GRCh38
rs137852916 Chr.6: 145635451-145635451 on Build GRCh38
rs755014724 Chr.6: 145635322-145635322 on Build GRCh38
rs371974399 Chr.6: 145635452-145635452 on Build GRCh38
rs781291421 Chr.6: 145635468-145635468 on Build GRCh38
rs750776112 Chr.6: 145635423-145635423 on Build GRCh38
rs367827948 Chr.6: 145635368-145635368 on Build GRCh38
rs796052423 Chr.6: 145635410-145635410 on Build GRCh38
rs777678187 Chr.6: 145635351-145635351 on Build GRCh38
rs748048885 Chr.6: 145635332-145635332 on Build GRCh38
rs796052435 Chr.6: 145635323-145635324 on Build GRCh38
rs777039727 Chr.6: 145635379-145635379 on Build GRCh38
rs966314930 Chr.6: 145635414-145635414 on Build GRCh38
rs142658692 Chr.6: 145635363-145635363 on Build GRCh38
rs191406622 Chr.6: 145635394-145635394 on Build GRCh38
rs778240670 Chr.6: 145635314-145635314 on Build GRCh38
rs1006950710 Chr.6: 145635303-145635303 on Build GRCh38
rs1045816 Chr.6: 145635435-145635435 on Build GRCh38
rs371088938 Chr.6: 145635370-145635370 on Build GRCh38
rs748995485 Chr.6: 145635360-145635360 on Build GRCh38
rs144565191 Chr.6: 145635319-145635319 on Build GRCh38
rs148475381 Chr.6: 145635331-145635331 on Build GRCh38
rs751742437 Chr.6: 145635462-145635462 on Build GRCh38

View Full Product Details