Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154575715-154575966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNB2
Entrez Gene ID
1141
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Gene Aliases
EFNL3, nAChRB2
UniGene
Hs.2306
Gene Chromosome Location
Chr.1: 154567781-154580026 on Build GRCh38
Transcripts
NM_000748.2 XM_017000180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
896633 c.1379G>A p.R460H Chr.1: 154575802-154575802 on Build GRCh38
2209200 c.1374C>T p.I458I Chr.1: 154575797-154575797 on Build GRCh38
6058381 c.1444C>G p.L482V Chr.1: 154575867-154575867 on Build GRCh38
6628770 c.1363G>A p.A455T Chr.1: 154575786-154575786 on Build GRCh38
7231032 c.1348G>A p.D450N Chr.1: 154575771-154575771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11803785 Chr.1: 154575739-154575739 on Build GRCh38
rs770206822 Chr.1: 154575748-154575757 on Build GRCh38
rs746029810 Chr.1: 154575768-154575768 on Build GRCh38
rs751272610 Chr.1: 154575795-154575795 on Build GRCh38
rs960649517 Chr.1: 154575900-154575900 on Build GRCh38
rs201110967 Chr.1: 154575776-154575776 on Build GRCh38
rs373985042 Chr.1: 154575803-154575803 on Build GRCh38
rs768798015 Chr.1: 154575920-154575920 on Build GRCh38
rs202079239 Chr.1: 154575801-154575801 on Build GRCh38
rs772270570 Chr.1: 154575836-154575836 on Build GRCh38
rs761717685 Chr.1: 154575782-154575782 on Build GRCh38
rs140505576 Chr.1: 154575798-154575798 on Build GRCh38
rs756629260 Chr.1: 154575747-154575747 on Build GRCh38
rs202135710 Chr.1: 154575846-154575846 on Build GRCh38
rs772921552 Chr.1: 154575785-154575785 on Build GRCh38
rs752248053 Chr.1: 154575861-154575861 on Build GRCh38
rs772146115 Chr.1: 154575775-154575775 on Build GRCh38
rs117167810 Chr.1: 154575746-154575746 on Build GRCh38
rs763453906 Chr.1: 154575879-154575879 on Build GRCh38
rs765202298 Chr.1: 154575783-154575783 on Build GRCh38
rs766229553 Chr.1: 154575791-154575791 on Build GRCh38
rs774575734 Chr.1: 154575766-154575766 on Build GRCh38
rs749633440 Chr.1: 154575762-154575762 on Build GRCh38
rs762839062 Chr.1: 154575787-154575787 on Build GRCh38
rs760253805 Chr.1: 154575875-154575875 on Build GRCh38
rs200177119 Chr.1: 154575756-154575756 on Build GRCh38
rs771240779 Chr.1: 154575764-154575764 on Build GRCh38
rs767243629 Chr.1: 154575857-154575857 on Build GRCh38
rs766283123 Chr.1: 154575850-154575850 on Build GRCh38
rs747080974 Chr.1: 154575845-154575845 on Build GRCh38
rs778371515 Chr.1: 154575818-154575818 on Build GRCh38
rs8192486 Chr.1: 154575905-154575905 on Build GRCh38
rs749294108 Chr.1: 154575945-154575945 on Build GRCh38
rs749637891 Chr.1: 154575827-154575832 on Build GRCh38
rs200582284 Chr.1: 154575911-154575911 on Build GRCh38
rs757759724 Chr.1: 154575890-154575890 on Build GRCh38
rs774272679 Chr.1: 154575853-154575853 on Build GRCh38
rs780794041 Chr.1: 154575743-154575743 on Build GRCh38
rs781068734 Chr.1: 154575931-154575931 on Build GRCh38
rs754372558 Chr.1: 154575889-154575889 on Build GRCh38
rs138886952 Chr.1: 154575830-154575830 on Build GRCh38
rs201578298 Chr.1: 154575738-154575738 on Build GRCh38
rs551094274 Chr.1: 154575810-154575810 on Build GRCh38
rs796052332 Chr.1: 154575830-154575833 on Build GRCh38
rs200467784 Chr.1: 154575849-154575849 on Build GRCh38
rs943465068 Chr.1: 154575745-154575745 on Build GRCh38
rs779309344 Chr.1: 154575892-154575892 on Build GRCh38
rs756619313 Chr.1: 154575809-154575809 on Build GRCh38
rs746157258 Chr.1: 154575834-154575834 on Build GRCh38
rs773226966 Chr.1: 154575848-154575848 on Build GRCh38
rs1052299583 Chr.1: 154575797-154575797 on Build GRCh38
rs79137415 Chr.1: 154575855-154575855 on Build GRCh38
rs200855905 Chr.1: 154575838-154575838 on Build GRCh38
rs144813907 Chr.1: 154575908-154575908 on Build GRCh38
rs749708943 Chr.1: 154575824-154575824 on Build GRCh38
rs747154910 Chr.1: 154575919-154575919 on Build GRCh38
rs778537596 Chr.1: 154575815-154575815 on Build GRCh38
rs200367867 Chr.1: 154575741-154575741 on Build GRCh38
rs796052328 Chr.1: 154575886-154575886 on Build GRCh38

View Full Product Details