Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 30905362-30905591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAN1
Entrez Gene ID
22909
Gene Name
FANCD2/FANCI-associated nuclease 1
Gene Aliases
KIAA1018, KMIN, MTMR15, hFAN1
UniGene
Hs.584863
Gene Chromosome Location
Chr.15: 30903695-30943108 on Build GRCh38
Transcripts
NM_001146094.1 NM_001146095.1 NM_001146096.1 NM_014967.4 XM_005254232.4 XM_005254234.4 XM_005254235.3 XM_011521370.2 XM_011521371.1 XM_011521372.2 XM_017022012.1 XM_017022013.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7256120 c.867C>T p.I289I Chr.15: 30905530-30905530 on Build GRCh38
2084288 c.775G>C p.D259H Chr.15: 30905438-30905438 on Build GRCh38
4510233 c.830C>T p.S277F Chr.15: 30905493-30905493 on Build GRCh38
1188942 c.918G>T p.M306I Chr.15: 30905581-30905581 on Build GRCh38
5215903 c.921_922delTG p.V308fs*5 Chr.15: 30905584-30905585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569290715 Chr.15: 30905488-30905488 on Build GRCh38
rs768645474 Chr.15: 30905545-30905545 on Build GRCh38
rs774210518 Chr.15: 30905398-30905398 on Build GRCh38
rs376256721 Chr.15: 30905499-30905499 on Build GRCh38
rs772129819 Chr.15: 30905420-30905420 on Build GRCh38
rs755602808 Chr.15: 30905458-30905458 on Build GRCh38
rs776738765 Chr.15: 30905431-30905431 on Build GRCh38
rs752301899 Chr.15: 30905456-30905456 on Build GRCh38
rs745804197 Chr.15: 30905521-30905521 on Build GRCh38
rs760772995 Chr.15: 30905425-30905425 on Build GRCh38
rs943955892 Chr.15: 30905408-30905408 on Build GRCh38
rs770745828 Chr.15: 30905395-30905395 on Build GRCh38
rs750964565 Chr.15: 30905500-30905500 on Build GRCh38
rs531416427 Chr.15: 30905479-30905479 on Build GRCh38
rs775709367 Chr.15: 30905422-30905422 on Build GRCh38
rs142437586 Chr.15: 30905446-30905446 on Build GRCh38
rs767736939 Chr.15: 30905387-30905387 on Build GRCh38
rs759975377 Chr.15: 30905531-30905533 on Build GRCh38
rs899678412 Chr.15: 30905443-30905443 on Build GRCh38
rs915072131 Chr.15: 30905484-30905484 on Build GRCh38
rs766861805 Chr.15: 30905562-30905562 on Build GRCh38
rs372989498 Chr.15: 30905434-30905434 on Build GRCh38
rs145950723 Chr.15: 30905550-30905550 on Build GRCh38
rs772275159 Chr.15: 30905544-30905544 on Build GRCh38
rs772091690 Chr.15: 30905525-30905525 on Build GRCh38
rs756991461 Chr.15: 30905496-30905496 on Build GRCh38
rs763532578 Chr.15: 30905559-30905559 on Build GRCh38
rs762225193 Chr.15: 30905435-30905435 on Build GRCh38
rs766931732 Chr.15: 30905452-30905452 on Build GRCh38
rs776844641 Chr.15: 30905549-30905549 on Build GRCh38
rs137920161 Chr.15: 30905445-30905445 on Build GRCh38
rs773780083 Chr.15: 30905555-30905555 on Build GRCh38
rs866963442 Chr.15: 30905502-30905502 on Build GRCh38
rs745822183 Chr.15: 30905399-30905399 on Build GRCh38
rs538297153 Chr.15: 30905528-30905528 on Build GRCh38
rs764422111 Chr.15: 30905427-30905427 on Build GRCh38
rs778740579 Chr.15: 30905509-30905509 on Build GRCh38
rs747058468 Chr.15: 30905540-30905540 on Build GRCh38
rs199810253 Chr.15: 30905470-30905470 on Build GRCh38
rs200798913 Chr.15: 30905394-30905394 on Build GRCh38

View Full Product Details