Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 227245861-227246100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A3
Entrez Gene ID
1285
Gene Name
collagen type IV alpha 3 chain
Gene Aliases
-
UniGene
Hs.570065
Gene Chromosome Location
Chr.2: 227164565-227314792 on Build GRCh38
Transcripts
NM_000091.4 XM_005246277.3 XM_005246280.3 XM_006712245.3 XM_011510555.1 XM_017003295.1 NR_102371.1

Gene Symbol
LOC654841
Entrez Gene ID
654841
Gene Name
uncharacterized LOC654841
Gene Aliases
-
UniGene
Hs.659982
Gene Chromosome Location
Chr.2: 227221052-227325164 on Build GRCh38
Transcripts
NM_000091.4 XM_005246277.3 XM_005246280.3 XM_006712245.3 XM_011510555.1 XM_017003295.1 NR_102371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4445699 c.325-3C>T p.? Chr.2: 227245951-227245951 on Build GRCh38
5861922 c.334G>A p.G112S Chr.2: 227245963-227245963 on Build GRCh38
4187061 c.326G>A p.G109D Chr.2: 227245955-227245955 on Build GRCh38
4631701 c.360C>T p.V120V Chr.2: 227245989-227245989 on Build GRCh38
5861921 c.334G>A p.G112S Chr.2: 227245963-227245963 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758630119 Chr.2: 227246006-227246006 on Build GRCh38
rs946624999 Chr.2: 227245926-227245926 on Build GRCh38
rs749455027 Chr.2: 227245919-227245919 on Build GRCh38
rs368386489 Chr.2: 227245928-227245928 on Build GRCh38
rs541435495 Chr.2: 227245910-227245910 on Build GRCh38
rs112712399 Chr.2: 227245927-227245927 on Build GRCh38
rs771264545 Chr.2: 227245980-227245981 on Build GRCh38
rs749696026 Chr.2: 227245971-227245971 on Build GRCh38
rs772238189 Chr.2: 227245930-227245930 on Build GRCh38
rs781264850 Chr.2: 227245918-227245918 on Build GRCh38
rs778886174 Chr.2: 227245990-227245990 on Build GRCh38
rs886055737 Chr.2: 227246015-227246015 on Build GRCh38
rs768591853 Chr.2: 227245923-227245923 on Build GRCh38
rs748188535 Chr.2: 227246000-227246000 on Build GRCh38
rs749390823 Chr.2: 227245972-227245972 on Build GRCh38
rs529104524 Chr.2: 227246038-227246038 on Build GRCh38
rs766935732 Chr.2: 227245994-227245994 on Build GRCh38
rs759125367 Chr.2: 227245937-227245937 on Build GRCh38
rs200381393 Chr.2: 227245917-227245917 on Build GRCh38
rs915040401 Chr.2: 227245925-227245925 on Build GRCh38
rs777955372 Chr.2: 227246033-227246033 on Build GRCh38
rs760968700 Chr.2: 227245934-227245934 on Build GRCh38
rs202147112 Chr.2: 227245972-227245972 on Build GRCh38
rs372029126 Chr.2: 227245932-227245932 on Build GRCh38
rs777841031 Chr.2: 227245961-227245964 on Build GRCh38
rs11680014 Chr.2: 227246077-227246077 on Build GRCh38
rs115324397 Chr.2: 227245975-227245975 on Build GRCh38
rs764631177 Chr.2: 227245939-227245939 on Build GRCh38
rs777117192 Chr.2: 227245936-227245936 on Build GRCh38
rs147052575 Chr.2: 227246046-227246046 on Build GRCh38
rs752211956 Chr.2: 227245949-227245949 on Build GRCh38
rs750308686 Chr.2: 227245980-227245980 on Build GRCh38
rs762518741 Chr.2: 227245953-227245953 on Build GRCh38
rs944334380 Chr.2: 227246073-227246073 on Build GRCh38
rs756033791 Chr.2: 227245989-227245989 on Build GRCh38
rs543390575 Chr.2: 227245958-227245958 on Build GRCh38
rs953599731 Chr.2: 227245979-227245979 on Build GRCh38

View Full Product Details