Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 75950686-75950941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPG1
Entrez Gene ID
3617
Gene Name
interphotoreceptor matrix proteoglycan 1
Gene Aliases
GP147, IPM150, SPACR, VMD4
UniGene
Hs.590893
Gene Chromosome Location
Chr.6: 75921114-76072678 on Build GRCh38
Transcripts
NM_001282368.1 NM_001563.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3175768 c.1627C>A p.P543T Chr.6: 75950759-75950759 on Build GRCh38
4438984 c.1597G>T p.V533L Chr.6: 75950789-75950789 on Build GRCh38
7200502 c.1555C>A p.L519I Chr.6: 75950831-75950831 on Build GRCh38
4914486 c.1638C>A p.F546L Chr.6: 75950748-75950748 on Build GRCh38
1697558 c.1550G>A p.R517K Chr.6: 75950836-75950836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780900706 Chr.6: 75950909-75950909 on Build GRCh38
rs760024120 Chr.6: 75950850-75950850 on Build GRCh38
rs763189552 Chr.6: 75950840-75950840 on Build GRCh38
rs201155813 Chr.6: 75950798-75950798 on Build GRCh38
rs773420926 Chr.6: 75950749-75950749 on Build GRCh38
rs754938809 Chr.6: 75950907-75950907 on Build GRCh38
rs756323714 Chr.6: 75950796-75950796 on Build GRCh38
rs963852045 Chr.6: 75950716-75950716 on Build GRCh38
rs767649818 Chr.6: 75950851-75950851 on Build GRCh38
rs779813555 Chr.6: 75950811-75950811 on Build GRCh38
rs368514950 Chr.6: 75950775-75950775 on Build GRCh38
rs976556833 Chr.6: 75950772-75950772 on Build GRCh38
rs775516577 Chr.6: 75950857-75950857 on Build GRCh38
rs1044618679 Chr.6: 75950731-75950731 on Build GRCh38
rs925970755 Chr.6: 75950876-75950876 on Build GRCh38
rs190423460 Chr.6: 75950725-75950725 on Build GRCh38
rs777226055 Chr.6: 75950917-75950917 on Build GRCh38
rs746517969 Chr.6: 75950823-75950823 on Build GRCh38
rs201558857 Chr.6: 75950808-75950808 on Build GRCh38
rs752974663 Chr.6: 75950788-75950788 on Build GRCh38
rs766748874 Chr.6: 75950758-75950758 on Build GRCh38
rs575857764 Chr.6: 75950914-75950914 on Build GRCh38
rs773650710 Chr.6: 75950838-75950838 on Build GRCh38
rs768219294 Chr.6: 75950825-75950825 on Build GRCh38
rs761005700 Chr.6: 75950859-75950859 on Build GRCh38
rs780849276 Chr.6: 75950829-75950829 on Build GRCh38
rs764171223 Chr.6: 75950866-75950866 on Build GRCh38
rs766350607 Chr.6: 75950872-75950872 on Build GRCh38
rs763567835 Chr.6: 75950753-75950753 on Build GRCh38
rs1029440187 Chr.6: 75950885-75950885 on Build GRCh38
rs780868773 Chr.6: 75950715-75950715 on Build GRCh38
rs774782339 Chr.6: 75950849-75950849 on Build GRCh38
rs376133758 Chr.6: 75950841-75950841 on Build GRCh38
rs367576664 Chr.6: 75950867-75950867 on Build GRCh38
rs747615571 Chr.6: 75950911-75950911 on Build GRCh38
rs79766540 Chr.6: 75950871-75950871 on Build GRCh38
rs35034020 Chr.6: 75950866-75950867 on Build GRCh38
rs558675396 Chr.6: 75950733-75950733 on Build GRCh38
rs3734311 Chr.6: 75950834-75950834 on Build GRCh38
rs751693422 Chr.6: 75950769-75950769 on Build GRCh38
rs751616903 Chr.6: 75950889-75950889 on Build GRCh38
rs759545902 Chr.6: 75950774-75950774 on Build GRCh38
rs750161331 Chr.6: 75950806-75950806 on Build GRCh38
rs768807327 Chr.6: 75950807-75950808 on Build GRCh38
rs747847816 Chr.6: 75950833-75950833 on Build GRCh38

View Full Product Details