Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 172427804-172428035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA6
Entrez Gene ID
3655
Gene Name
integrin subunit alpha 6
Gene Aliases
CD49f, ITGA6B, VLA-6
UniGene
Hs.133397
Gene Chromosome Location
Chr.2: 172427354-172506455 on Build GRCh38
Transcripts
NM_000210.3 NM_001079818.2 NM_001316306.1 XM_006712510.1 XM_006712511.1 XM_017004005.1 XM_017004006.1 XM_017004007.1 XM_017004008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660056 c.44G>T p.G15V Chr.2: 172427832-172427832 on Build GRCh38
5666562 c.96C>G p.N32K Chr.2: 172427884-172427884 on Build GRCh38
3372481 c.138C>T p.F46F Chr.2: 172427926-172427926 on Build GRCh38
6036586 c.96C>T p.N32N Chr.2: 172427884-172427884 on Build GRCh38
7372569 c.157C>G p.Q53E Chr.2: 172427945-172427945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376400633 Chr.2: 172427854-172427854 on Build GRCh38
rs749534380 Chr.2: 172427875-172427875 on Build GRCh38
rs776183303 Chr.2: 172427905-172427905 on Build GRCh38
rs374272135 Chr.2: 172427945-172427945 on Build GRCh38
rs754921421 Chr.2: 172428019-172428019 on Build GRCh38
rs6744873 Chr.2: 172427981-172427981 on Build GRCh38
rs965204894 Chr.2: 172427885-172427885 on Build GRCh38
rs746942286 Chr.2: 172427845-172427845 on Build GRCh38
rs755847509 Chr.2: 172427956-172427956 on Build GRCh38
rs982706769 Chr.2: 172428008-172428008 on Build GRCh38
rs746019316 Chr.2: 172427897-172427897 on Build GRCh38
rs752456003 Chr.2: 172427899-172427899 on Build GRCh38
rs138572695 Chr.2: 172427936-172427936 on Build GRCh38
rs1006950275 Chr.2: 172427878-172427878 on Build GRCh38
rs377085042 Chr.2: 172427926-172427926 on Build GRCh38
rs150472149 Chr.2: 172427843-172427843 on Build GRCh38
rs779259818 Chr.2: 172427962-172427962 on Build GRCh38
rs758278211 Chr.2: 172427829-172427829 on Build GRCh38
rs751546962 Chr.2: 172427938-172427938 on Build GRCh38
rs765517528 Chr.2: 172427999-172427999 on Build GRCh38
rs777091551 Chr.2: 172427915-172427915 on Build GRCh38
rs747879718 Chr.2: 172427953-172427953 on Build GRCh38
rs765897110 Chr.2: 172428009-172428009 on Build GRCh38
rs375279228 Chr.2: 172427996-172427996 on Build GRCh38
rs564206622 Chr.2: 172427826-172427826 on Build GRCh38
rs972560376 Chr.2: 172427888-172427888 on Build GRCh38
rs761372308 Chr.2: 172427923-172427923 on Build GRCh38
rs767272243 Chr.2: 172427834-172427834 on Build GRCh38
rs750028773 Chr.2: 172427934-172427934 on Build GRCh38
rs771118805 Chr.2: 172427890-172427890 on Build GRCh38
rs6716540 Chr.2: 172427985-172427985 on Build GRCh38
rs758359210 Chr.2: 172427970-172427971 on Build GRCh38
rs548107179 Chr.2: 172427997-172427997 on Build GRCh38
rs1020126767 Chr.2: 172427883-172427883 on Build GRCh38
rs13009675 Chr.2: 172427982-172427982 on Build GRCh38
rs780969691 Chr.2: 172427952-172427952 on Build GRCh38
rs761245728 Chr.2: 172427907-172427907 on Build GRCh38
rs6731763 Chr.2: 172428000-172428000 on Build GRCh38
rs140763977 Chr.2: 172427848-172427848 on Build GRCh38
rs954207601 Chr.2: 172427864-172427864 on Build GRCh38
rs533391020 Chr.2: 172428006-172428006 on Build GRCh38
rs544002743 Chr.2: 172427937-172427937 on Build GRCh38
rs774309985 Chr.2: 172427894-172427894 on Build GRCh38

View Full Product Details