Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150520297-150520522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP7
Entrez Gene ID
168537
Gene Name
GTPase, IMAP family member 7
Gene Aliases
IAN7, hIAN7
UniGene
Hs.647074
Gene Chromosome Location
Chr.7: 150514857-150521073 on Build GRCh38
Transcripts
NM_153236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5669204 c.375G>C p.K125N Chr.7: 150520349-150520349 on Build GRCh38
4437499 c.448G>A p.D150N Chr.7: 150520422-150520422 on Build GRCh38
3923080 c.414G>T p.E138D Chr.7: 150520388-150520388 on Build GRCh38
1087231 c.503G>A p.S168N Chr.7: 150520477-150520477 on Build GRCh38
5358268 c.334G>A p.V112I Chr.7: 150520308-150520308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530097290 Chr.7: 150520360-150520360 on Build GRCh38
rs1010189661 Chr.7: 150520319-150520319 on Build GRCh38
rs770777133 Chr.7: 150520451-150520451 on Build GRCh38
rs201058399 Chr.7: 150520371-150520371 on Build GRCh38
rs916753968 Chr.7: 150520395-150520395 on Build GRCh38
rs910145240 Chr.7: 150520490-150520490 on Build GRCh38
rs757301966 Chr.7: 150520383-150520383 on Build GRCh38
rs115164158 Chr.7: 150520353-150520353 on Build GRCh38
rs80028286 Chr.7: 150520495-150520495 on Build GRCh38
rs746883768 Chr.7: 150520446-150520446 on Build GRCh38
rs771759006 Chr.7: 150520355-150520355 on Build GRCh38
rs755433641 Chr.7: 150520433-150520433 on Build GRCh38
rs116511786 Chr.7: 150520434-150520434 on Build GRCh38
rs764216565 Chr.7: 150520377-150520377 on Build GRCh38
rs992435888 Chr.7: 150520389-150520389 on Build GRCh38
rs773775975 Chr.7: 150520430-150520430 on Build GRCh38
rs747843953 Chr.7: 150520438-150520438 on Build GRCh38
rs769675039 Chr.7: 150520470-150520470 on Build GRCh38
rs759238652 Chr.7: 150520461-150520461 on Build GRCh38
rs751865182 Chr.7: 150520382-150520382 on Build GRCh38
rs765040620 Chr.7: 150520455-150520455 on Build GRCh38
rs747618380 Chr.7: 150520477-150520478 on Build GRCh38
rs141641243 Chr.7: 150520369-150520369 on Build GRCh38
rs774522502 Chr.7: 150520483-150520483 on Build GRCh38
rs146170876 Chr.7: 150520429-150520429 on Build GRCh38
rs780402746 Chr.7: 150520421-150520421 on Build GRCh38
rs762049978 Chr.7: 150520489-150520489 on Build GRCh38
rs763167681 Chr.7: 150520372-150520372 on Build GRCh38
rs369222429 Chr.7: 150520331-150520331 on Build GRCh38
rs767533437 Chr.7: 150520497-150520497 on Build GRCh38
rs900606701 Chr.7: 150520319-150520326 on Build GRCh38
rs767621267 Chr.7: 150520415-150520415 on Build GRCh38
rs771669495 Chr.7: 150520442-150520442 on Build GRCh38
rs773254207 Chr.7: 150520364-150520364 on Build GRCh38
rs928288537 Chr.7: 150520437-150520437 on Build GRCh38
rs750886687 Chr.7: 150520419-150520419 on Build GRCh38
rs756630060 Chr.7: 150520420-150520420 on Build GRCh38
rs373463019 Chr.7: 150520376-150520376 on Build GRCh38
rs749614181 Chr.7: 150520425-150520425 on Build GRCh38
rs750508828 Chr.7: 150520500-150520500 on Build GRCh38

View Full Product Details