Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128264538-128264805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEFSEC
Entrez Gene ID
60678
Gene Name
eukaryotic elongation factor, selenocysteine-tRNA specific
Gene Aliases
EFSEC, SELB
UniGene
Hs.477498
Gene Chromosome Location
Chr.3: 128153456-128426193 on Build GRCh38
Transcripts
NM_021937.4 XM_005247696.2 XM_011513066.2 XM_017006975.1 XM_017006976.1 XM_017006977.1 XM_017006978.1 XM_017006979.1 XM_017006980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4113245 c.629C>T p.T210M Chr.3: 128264624-128264624 on Build GRCh38
4113246 c.721A>G p.M241V Chr.3: 128264716-128264716 on Build GRCh38
3059479 c.656A>G p.D219G Chr.3: 128264651-128264651 on Build GRCh38
4503709 c.646C>T p.P216S Chr.3: 128264641-128264641 on Build GRCh38
1495386 c.689A>T p.H230L Chr.3: 128264684-128264684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374217170 Chr.3: 128264591-128264591 on Build GRCh38
rs769621965 Chr.3: 128264750-128264751 on Build GRCh38
rs747933691 Chr.3: 128264652-128264652 on Build GRCh38
rs371163912 Chr.3: 128264587-128264587 on Build GRCh38
rs774900953 Chr.3: 128264667-128264667 on Build GRCh38
rs148854623 Chr.3: 128264658-128264658 on Build GRCh38
rs753789877 Chr.3: 128264645-128264645 on Build GRCh38
rs151307884 Chr.3: 128264751-128264751 on Build GRCh38
rs765067707 Chr.3: 128264648-128264648 on Build GRCh38
rs987125505 Chr.3: 128264586-128264586 on Build GRCh38
rs2811544 Chr.3: 128264652-128264652 on Build GRCh38
rs759747927 Chr.3: 128264734-128264734 on Build GRCh38
rs766592377 Chr.3: 128264640-128264640 on Build GRCh38
rs758108228 Chr.3: 128264745-128264745 on Build GRCh38
rs780879674 Chr.3: 128264763-128264763 on Build GRCh38
rs371605557 Chr.3: 128264646-128264646 on Build GRCh38
rs1020162992 Chr.3: 128264583-128264583 on Build GRCh38
rs760646864 Chr.3: 128264631-128264631 on Build GRCh38
rs367956823 Chr.3: 128264657-128264657 on Build GRCh38
rs760991291 Chr.3: 128264698-128264698 on Build GRCh38
rs758282576 Chr.3: 128264653-128264653 on Build GRCh38
rs929763108 Chr.3: 128264644-128264644 on Build GRCh38
rs749928084 Chr.3: 128264766-128264766 on Build GRCh38
rs773260814 Chr.3: 128264687-128264687 on Build GRCh38
rs200179846 Chr.3: 128264677-128264677 on Build GRCh38
rs192426318 Chr.3: 128264625-128264625 on Build GRCh38
rs769134730 Chr.3: 128264666-128264666 on Build GRCh38
rs374856982 Chr.3: 128264712-128264712 on Build GRCh38
rs780685267 Chr.3: 128264664-128264664 on Build GRCh38
rs142548867 Chr.3: 128264663-128264663 on Build GRCh38
rs755656837 Chr.3: 128264770-128264770 on Build GRCh38
rs955284091 Chr.3: 128264584-128264584 on Build GRCh38
rs578191693 Chr.3: 128264740-128264740 on Build GRCh38
rs766504562 Chr.3: 128264709-128264709 on Build GRCh38
rs867798525 Chr.3: 128264769-128264769 on Build GRCh38
rs990911210 Chr.3: 128264741-128264741 on Build GRCh38
rs752722886 Chr.3: 128264742-128264742 on Build GRCh38
rs751104507 Chr.3: 128264752-128264752 on Build GRCh38
rs749823720 Chr.3: 128264588-128264588 on Build GRCh38
rs748709387 Chr.3: 128264775-128264775 on Build GRCh38
rs756915873 Chr.3: 128264761-128264761 on Build GRCh38
rs551355460 Chr.3: 128264563-128264563 on Build GRCh38
rs774710839 Chr.3: 128264604-128264604 on Build GRCh38
rs773521566 Chr.3: 128264628-128264628 on Build GRCh38

View Full Product Details