Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157675013-157675280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf52
Entrez Gene ID
100190949
Gene Name
chromosome 5 open reading frame 52
Gene Aliases
-
UniGene
Hs.720862
Gene Chromosome Location
Chr.5: 157671237-157680176 on Build GRCh38
Transcripts
NM_001145132.1 XM_011534416.1 XM_017008909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5835002 c.316_317insAC p.M106fs*4 Chr.5: 157675195-157675196 on Build GRCh38
7358618 c.236C>T p.A79V Chr.5: 157675115-157675115 on Build GRCh38
7216581 c.272G>A p.R91Q Chr.5: 157675151-157675151 on Build GRCh38
5835001 c.314_315insCTT p.E105>DL Chr.5: 157675193-157675194 on Build GRCh38
4442192 c.235G>T p.A79S Chr.5: 157675114-157675114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024615316 Chr.5: 157675139-157675139 on Build GRCh38
rs550237548 Chr.5: 157675201-157675201 on Build GRCh38
rs769691822 Chr.5: 157675205-157675205 on Build GRCh38
rs142164982 Chr.5: 157675115-157675115 on Build GRCh38
rs988266568 Chr.5: 157675176-157675176 on Build GRCh38
rs769593871 Chr.5: 157675048-157675048 on Build GRCh38
rs747988766 Chr.5: 157675202-157675202 on Build GRCh38
rs760989660 Chr.5: 157675240-157675240 on Build GRCh38
rs10051782 Chr.5: 157675055-157675055 on Build GRCh38
rs565344193 Chr.5: 157675120-157675120 on Build GRCh38
rs964493376 Chr.5: 157675183-157675183 on Build GRCh38
rs776371736 Chr.5: 157675054-157675054 on Build GRCh38
rs545174583 Chr.5: 157675172-157675172 on Build GRCh38
rs746684194 Chr.5: 157675196-157675196 on Build GRCh38
rs546397461 Chr.5: 157675125-157675125 on Build GRCh38
rs200431937 Chr.5: 157675171-157675171 on Build GRCh38
rs10051838 Chr.5: 157675151-157675151 on Build GRCh38
rs760891521 Chr.5: 157675141-157675141 on Build GRCh38
rs762308596 Chr.5: 157675247-157675247 on Build GRCh38
rs750582565 Chr.5: 157675223-157675223 on Build GRCh38
rs559959230 Chr.5: 157675134-157675134 on Build GRCh38
rs78469140 Chr.5: 157675148-157675148 on Build GRCh38
rs540586017 Chr.5: 157675077-157675077 on Build GRCh38
rs754258297 Chr.5: 157675153-157675153 on Build GRCh38
rs768550713 Chr.5: 157675049-157675049 on Build GRCh38
rs759599069 Chr.5: 157675206-157675206 on Build GRCh38
rs752388538 Chr.5: 157675037-157675038 on Build GRCh38
rs772975944 Chr.5: 157675149-157675152 on Build GRCh38
rs893502122 Chr.5: 157675136-157675136 on Build GRCh38
rs777030657 Chr.5: 157675246-157675246 on Build GRCh38
rs6884959 Chr.5: 157675177-157675177 on Build GRCh38
rs775509616 Chr.5: 157675235-157675235 on Build GRCh38
rs560695628 Chr.5: 157675211-157675211 on Build GRCh38
rs764314650 Chr.5: 157675245-157675245 on Build GRCh38
rs564739954 Chr.5: 157675175-157675175 on Build GRCh38
rs779555624 Chr.5: 157675184-157675184 on Build GRCh38
rs191694326 Chr.5: 157675118-157675118 on Build GRCh38
rs760651792 Chr.5: 157675245-157675256 on Build GRCh38
rs996302084 Chr.5: 157675215-157675215 on Build GRCh38
rs749293588 Chr.5: 157675064-157675064 on Build GRCh38
rs983819411 Chr.5: 157675250-157675250 on Build GRCh38

View Full Product Details