Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3885675-3885942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATCAY
Entrez Gene ID
85300
Gene Name
ATCAY, caytaxin
Gene Aliases
BNIP-H, CLAC
UniGene
Hs.418055
Gene Chromosome Location
Chr.19: 3880620-3928082 on Build GRCh38
Transcripts
NM_033064.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3388946 c.42C>T p.D14D Chr.19: 3885809-3885809 on Build GRCh38
5389639 c.74C>T p.P25L Chr.19: 3885841-3885841 on Build GRCh38
1183869 c.37G>A p.V13M Chr.19: 3885804-3885804 on Build GRCh38
1183868 c.37G>A p.V13M Chr.19: 3885804-3885804 on Build GRCh38
3202147 c.51G>C p.E17D Chr.19: 3885818-3885818 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745720974 Chr.19: 3885804-3885804 on Build GRCh38
rs74912608 Chr.19: 3885920-3885920 on Build GRCh38
rs968674495 Chr.19: 3885895-3885895 on Build GRCh38
rs375170455 Chr.19: 3885885-3885885 on Build GRCh38
rs868017605 Chr.19: 3885780-3885780 on Build GRCh38
rs765457963 Chr.19: 3885877-3885877 on Build GRCh38
rs942528475 Chr.19: 3885741-3885741 on Build GRCh38
rs529428286 Chr.19: 3885714-3885714 on Build GRCh38
rs978384357 Chr.19: 3885918-3885918 on Build GRCh38
rs56964691 Chr.19: 3885883-3885883 on Build GRCh38
rs968800488 Chr.19: 3885703-3885703 on Build GRCh38
rs748431773 Chr.19: 3885788-3885788 on Build GRCh38
rs760237730 Chr.19: 3885857-3885857 on Build GRCh38
rs1048180602 Chr.19: 3885880-3885880 on Build GRCh38
rs1057024264 Chr.19: 3885787-3885787 on Build GRCh38
rs534896329 Chr.19: 3885873-3885873 on Build GRCh38
rs937126056 Chr.19: 3885778-3885778 on Build GRCh38
rs775624223 Chr.19: 3885810-3885810 on Build GRCh38
rs374309679 Chr.19: 3885879-3885879 on Build GRCh38
rs772127259 Chr.19: 3885809-3885809 on Build GRCh38
rs368213837 Chr.19: 3885803-3885803 on Build GRCh38
rs73919386 Chr.19: 3885708-3885708 on Build GRCh38
rs761543482 Chr.19: 3885876-3885876 on Build GRCh38
rs146935412 Chr.19: 3885713-3885713 on Build GRCh38
rs768315218 Chr.19: 3885861-3885861 on Build GRCh38
rs1037918542 Chr.19: 3885894-3885894 on Build GRCh38
rs770582696 Chr.19: 3885793-3885793 on Build GRCh38
rs927203522 Chr.19: 3885768-3885768 on Build GRCh38
rs565788691 Chr.19: 3885830-3885830 on Build GRCh38
rs1022486334 Chr.19: 3885886-3885886 on Build GRCh38

View Full Product Details