Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 32093236-32093461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP5
Entrez Gene ID
394
Gene Name
Rho GTPase activating protein 5
Gene Aliases
GFI2, RhoGAP5, p190-B, p190BRhoGAP
UniGene
Hs.592313
Gene Chromosome Location
Chr.14: 32076152-32159728 on Build GRCh38
Transcripts
NM_001030055.1 NM_001173.2 XM_005267635.3 XM_005267636.3 XM_017021287.1 XM_017021288.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6603672 c.2575C>T p.R859W Chr.14: 32093244-32093244 on Build GRCh38
6880270 c.2602G>A p.A868T Chr.14: 32093271-32093271 on Build GRCh38
6075939 c.2591G>T p.G864V Chr.14: 32093260-32093260 on Build GRCh38
955341 c.2759C>T p.S920F Chr.14: 32093428-32093428 on Build GRCh38
256146 c.2669A>T p.D890V Chr.14: 32093338-32093338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925999127 Chr.14: 32093360-32093360 on Build GRCh38
rs759522421 Chr.14: 32093386-32093386 on Build GRCh38
rs770785852 Chr.14: 32093378-32093378 on Build GRCh38
rs143191152 Chr.14: 32093363-32093363 on Build GRCh38
rs761990330 Chr.14: 32093432-32093432 on Build GRCh38
rs145372823 Chr.14: 32093417-32093417 on Build GRCh38
rs368445681 Chr.14: 32093385-32093385 on Build GRCh38
rs747786675 Chr.14: 32093327-32093327 on Build GRCh38
rs780375788 Chr.14: 32093297-32093297 on Build GRCh38
rs200739729 Chr.14: 32093279-32093279 on Build GRCh38
rs61737869 Chr.14: 32093303-32093303 on Build GRCh38
rs781483073 Chr.14: 32093339-32093339 on Build GRCh38
rs557284303 Chr.14: 32093273-32093273 on Build GRCh38
rs754375160 Chr.14: 32093306-32093306 on Build GRCh38
rs940479761 Chr.14: 32093295-32093296 on Build GRCh38
rs757485463 Chr.14: 32093269-32093269 on Build GRCh38
rs150461143 Chr.14: 32093370-32093370 on Build GRCh38
rs749147518 Chr.14: 32093375-32093375 on Build GRCh38
rs764381821 Chr.14: 32093361-32093361 on Build GRCh38
rs780764419 Chr.14: 32093324-32093324 on Build GRCh38
rs147957990 Chr.14: 32093291-32093291 on Build GRCh38

View Full Product Details