Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196507814-196508043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMCO1
Entrez Gene ID
255798
Gene Name
single-pass membrane protein with coiled-coil domains 1
Gene Aliases
C3orf43
UniGene
Hs.631933
Gene Chromosome Location
Chr.3: 196506877-196515366 on Build GRCh38
Transcripts
NM_001077657.2 NM_001320473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
230025 c.627A>G p.I209M Chr.3: 196507905-196507905 on Build GRCh38
5769995 c.615C>T p.L205L Chr.3: 196507917-196507917 on Build GRCh38
7177369 c.568G>C p.E190Q Chr.3: 196507964-196507964 on Build GRCh38
3847159 c.576G>T p.G192G Chr.3: 196507956-196507956 on Build GRCh38
2947359 c.550A>G p.R184G Chr.3: 196507982-196507982 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760303135 Chr.3: 196507857-196507857 on Build GRCh38
rs62410569 Chr.3: 196507916-196507916 on Build GRCh38
rs749575279 Chr.3: 196507950-196507950 on Build GRCh38
rs781291320 Chr.3: 196507917-196507917 on Build GRCh38
rs927456808 Chr.3: 196508009-196508009 on Build GRCh38
rs371825571 Chr.3: 196507966-196507966 on Build GRCh38
rs374565107 Chr.3: 196508003-196508003 on Build GRCh38
rs192469355 Chr.3: 196507997-196507997 on Build GRCh38
rs111869058 Chr.3: 196507844-196507844 on Build GRCh38
rs778331783 Chr.3: 196507968-196507969 on Build GRCh38
rs116717850 Chr.3: 196507976-196507976 on Build GRCh38
rs964921644 Chr.3: 196507996-196507996 on Build GRCh38
rs753763653 Chr.3: 196507903-196507903 on Build GRCh38
rs201420620 Chr.3: 196507965-196507965 on Build GRCh38
rs560601943 Chr.3: 196507842-196507842 on Build GRCh38
rs368493245 Chr.3: 196507879-196507879 on Build GRCh38
rs368414591 Chr.3: 196507854-196507854 on Build GRCh38
rs571248406 Chr.3: 196507885-196507885 on Build GRCh38
rs187809363 Chr.3: 196507906-196507906 on Build GRCh38
rs370580225 Chr.3: 196507951-196507951 on Build GRCh38
rs376283410 Chr.3: 196507910-196507910 on Build GRCh38
rs367904648 Chr.3: 196507940-196507940 on Build GRCh38
rs763818619 Chr.3: 196507884-196507884 on Build GRCh38
rs777955000 Chr.3: 196507943-196507943 on Build GRCh38
rs769907990 Chr.3: 196507942-196507942 on Build GRCh38
rs546274858 Chr.3: 196507874-196507874 on Build GRCh38
rs774339055 Chr.3: 196507978-196507978 on Build GRCh38
rs757252804 Chr.3: 196507932-196507933 on Build GRCh38
rs780285836 Chr.3: 196507903-196507903 on Build GRCh38
rs771876680 Chr.3: 196507969-196507969 on Build GRCh38

View Full Product Details