Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120072570-120072805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICDL1
Entrez Gene ID
92558
Gene Name
BICD family like cargo adaptor 1
Gene Aliases
BICDR-1, BICDR1, CCDC64, H_267D11.1
UniGene
Hs.369763
Gene Chromosome Location
Chr.12: 119989806-120094495 on Build GRCh38
Transcripts
NM_207311.2 XM_005253991.4 XM_006719694.3 XM_006719696.3 XM_006719697.3 XM_011538999.2 XM_011539000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3772285 c.183C>T p.A61A Chr.12: 120072657-120072657 on Build GRCh38
4461435 c.1205C>T p.S402L Chr.12: 120072626-120072626 on Build GRCh38
1510944 c.1214C>T p.S405L Chr.12: 120072635-120072635 on Build GRCh38
317094 c.1247G>T p.R416L Chr.12: 120072668-120072668 on Build GRCh38
5657052 c.194G>T p.R65L Chr.12: 120072668-120072668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372129224 Chr.12: 120072613-120072613 on Build GRCh38
rs199508010 Chr.12: 120072668-120072668 on Build GRCh38
rs757654492 Chr.12: 120072680-120072680 on Build GRCh38
rs374769468 Chr.12: 120072635-120072635 on Build GRCh38
rs754241625 Chr.12: 120072652-120072652 on Build GRCh38
rs879706173 Chr.12: 120072758-120072758 on Build GRCh38
rs376828634 Chr.12: 120072594-120072594 on Build GRCh38
rs779009135 Chr.12: 120072658-120072658 on Build GRCh38
rs760026562 Chr.12: 120072609-120072609 on Build GRCh38
rs1033869619 Chr.12: 120072729-120072729 on Build GRCh38
rs369210438 Chr.12: 120072657-120072657 on Build GRCh38
rs764700583 Chr.12: 120072758-120072758 on Build GRCh38
rs368184092 Chr.12: 120072595-120072595 on Build GRCh38
rs770659165 Chr.12: 120072761-120072762 on Build GRCh38
rs776176455 Chr.12: 120072748-120072748 on Build GRCh38
rs762432766 Chr.12: 120072738-120072738 on Build GRCh38
rs200012706 Chr.12: 120072602-120072602 on Build GRCh38
rs990020306 Chr.12: 120072779-120072779 on Build GRCh38
rs539360773 Chr.12: 120072782-120072782 on Build GRCh38
rs570588809 Chr.12: 120072740-120072740 on Build GRCh38
rs758297012 Chr.12: 120072661-120072661 on Build GRCh38
rs981782062 Chr.12: 120072725-120072725 on Build GRCh38
rs369475993 Chr.12: 120072687-120072687 on Build GRCh38
rs375531501 Chr.12: 120072675-120072675 on Build GRCh38
rs772724969 Chr.12: 120072707-120072707 on Build GRCh38
rs539893870 Chr.12: 120072766-120072766 on Build GRCh38
rs764503470 Chr.12: 120072768-120072768 on Build GRCh38
rs201940570 Chr.12: 120072667-120072667 on Build GRCh38
rs550694789 Chr.12: 120072659-120072659 on Build GRCh38
rs756610038 Chr.12: 120072636-120072636 on Build GRCh38
rs769338259 Chr.12: 120072692-120072692 on Build GRCh38
rs746776288 Chr.12: 120072669-120072669 on Build GRCh38

View Full Product Details