Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143726325-143726570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83H
Entrez Gene ID
286077
Gene Name
family with sequence similarity 83 member H
Gene Aliases
AI3
UniGene
Hs.67776
Gene Chromosome Location
Chr.8: 143723933-143739947 on Build GRCh38
Transcripts
NM_198488.3 XM_005250887.3 XM_005250888.3 XM_005250889.3 XM_011516980.2 XM_011516981.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6530559 c.3130C>G p.L1044V Chr.8: 143726331-143726331 on Build GRCh38
3412808 c.3002G>A p.R1001H Chr.8: 143726459-143726459 on Build GRCh38
1161742 c.2998C>T p.R1000W Chr.8: 143726463-143726463 on Build GRCh38
5821725 c.2910G>T p.R970S Chr.8: 143726551-143726551 on Build GRCh38
1733306 c.2980G>A p.G994S Chr.8: 143726481-143726481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781897628 Chr.8: 143726460-143726460 on Build GRCh38
rs782660797 Chr.8: 143726514-143726514 on Build GRCh38
rs201196465 Chr.8: 143726492-143726492 on Build GRCh38
rs782413434 Chr.8: 143726545-143726545 on Build GRCh38
rs782740837 Chr.8: 143726395-143726395 on Build GRCh38
rs782717184 Chr.8: 143726387-143726387 on Build GRCh38
rs781961685 Chr.8: 143726400-143726400 on Build GRCh38
rs578053048 Chr.8: 143726381-143726381 on Build GRCh38
rs782294315 Chr.8: 143726410-143726410 on Build GRCh38
rs782162013 Chr.8: 143726538-143726538 on Build GRCh38
rs376662947 Chr.8: 143726404-143726404 on Build GRCh38
rs782421765 Chr.8: 143726412-143726412 on Build GRCh38
rs532022507 Chr.8: 143726493-143726493 on Build GRCh38
rs536891981 Chr.8: 143726362-143726362 on Build GRCh38
rs201981905 Chr.8: 143726518-143726518 on Build GRCh38
rs782748080 Chr.8: 143726481-143726481 on Build GRCh38
rs376168041 Chr.8: 143726527-143726527 on Build GRCh38
rs369700124 Chr.8: 143726400-143726400 on Build GRCh38
rs781992350 Chr.8: 143726408-143726408 on Build GRCh38
rs117987215 Chr.8: 143726402-143726402 on Build GRCh38
rs970556209 Chr.8: 143726371-143726371 on Build GRCh38
rs369951434 Chr.8: 143726406-143726406 on Build GRCh38
rs372205376 Chr.8: 143726390-143726390 on Build GRCh38
rs542994892 Chr.8: 143726463-143726463 on Build GRCh38
rs782720254 Chr.8: 143726464-143726464 on Build GRCh38
rs781980123 Chr.8: 143726535-143726535 on Build GRCh38
rs112740523 Chr.8: 143726344-143726344 on Build GRCh38
rs376189581 Chr.8: 143726482-143726482 on Build GRCh38
rs781784657 Chr.8: 143726466-143726466 on Build GRCh38
rs781958263 Chr.8: 143726364-143726364 on Build GRCh38
rs943655969 Chr.8: 143726487-143726487 on Build GRCh38
rs782223642 Chr.8: 143726405-143726405 on Build GRCh38
rs372968379 Chr.8: 143726431-143726431 on Build GRCh38
rs191891370 Chr.8: 143726500-143726500 on Build GRCh38
rs781949444 Chr.8: 143726441-143726441 on Build GRCh38
rs929013671 Chr.8: 143726511-143726511 on Build GRCh38
rs200636666 Chr.8: 143726523-143726523 on Build GRCh38
rs768891623 Chr.8: 143726374-143726374 on Build GRCh38
rs781935067 Chr.8: 143726422-143726422 on Build GRCh38
rs782036749 Chr.8: 143726378-143726378 on Build GRCh38
rs761661915 Chr.8: 143726516-143726516 on Build GRCh38
rs781790060 Chr.8: 143726389-143726389 on Build GRCh38
rs377063712 Chr.8: 143726394-143726394 on Build GRCh38
rs781928465 Chr.8: 143726548-143726548 on Build GRCh38
rs782129198 Chr.8: 143726452-143726452 on Build GRCh38
rs910789209 Chr.8: 143726484-143726484 on Build GRCh38
rs782745300 Chr.8: 143726436-143726436 on Build GRCh38
rs782084718 Chr.8: 143726470-143726470 on Build GRCh38
rs781867409 Chr.8: 143726397-143726397 on Build GRCh38
rs782214215 Chr.8: 143726525-143726525 on Build GRCh38
rs782507248 Chr.8: 143726398-143726398 on Build GRCh38

View Full Product Details