Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128946394-128946639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRF5
Entrez Gene ID
3663
Gene Name
interferon regulatory factor 5
Gene Aliases
SLEB10
UniGene
Hs.521181
Gene Chromosome Location
Chr.7: 128937737-128950042 on Build GRCh38
Transcripts
NM_001098627.3 NM_001098629.2 NM_001098630.2 NM_001242452.2 NM_032643.4 XM_005250317.3 XM_006715974.2 XM_011516158.2 XM_011516159.2 XM_011516160.1 XM_011516161.1 XM_011516162.1 XM_011516163.2 XM_011516164.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5113952 c.393_394insC p.E134fs*1 Chr.7: 128946508-128946509 on Build GRCh38
6548742 c.394C>T p.P132S Chr.7: 128946509-128946509 on Build GRCh38
5952613 c.430G>A p.E144K Chr.7: 128946545-128946545 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776036438 Chr.7: 128946593-128946593 on Build GRCh38
rs201418545 Chr.7: 128946562-128946562 on Build GRCh38
rs766371118 Chr.7: 128946616-128946616 on Build GRCh38
rs531640976 Chr.7: 128946425-128946425 on Build GRCh38
rs779832149 Chr.7: 128946498-128946498 on Build GRCh38
rs935497546 Chr.7: 128946434-128946435 on Build GRCh38
rs371335617 Chr.7: 128946556-128946556 on Build GRCh38
rs776966512 Chr.7: 128946567-128946567 on Build GRCh38
rs200812729 Chr.7: 128946600-128946600 on Build GRCh38
rs377609927 Chr.7: 128946578-128946578 on Build GRCh38
rs756115795 Chr.7: 128946496-128946496 on Build GRCh38
rs150186560 Chr.7: 128946454-128946454 on Build GRCh38
rs749950026 Chr.7: 128946490-128946490 on Build GRCh38
rs772266449 Chr.7: 128946536-128946536 on Build GRCh38
rs775238687 Chr.7: 128946606-128946607 on Build GRCh38
rs761626157 Chr.7: 128946599-128946599 on Build GRCh38
rs767194622 Chr.7: 128946478-128946478 on Build GRCh38
rs373171551 Chr.7: 128946569-128946569 on Build GRCh38
rs35062949 Chr.7: 128946466-128946467 on Build GRCh38
rs779108354 Chr.7: 128946512-128946512 on Build GRCh38
rs371885677 Chr.7: 128946459-128946459 on Build GRCh38
rs367670808 Chr.7: 128946438-128946438 on Build GRCh38
rs943756715 Chr.7: 128946528-128946528 on Build GRCh38
rs80269234 Chr.7: 128946428-128946428 on Build GRCh38
rs773344520 Chr.7: 128946543-128946543 on Build GRCh38
rs748307649 Chr.7: 128946532-128946532 on Build GRCh38
rs779736518 Chr.7: 128946620-128946620 on Build GRCh38
rs765639543 Chr.7: 128946572-128946572 on Build GRCh38
rs749129718 Chr.7: 128946509-128946509 on Build GRCh38
rs1048732408 Chr.7: 128946621-128946621 on Build GRCh38
rs755876932 Chr.7: 128946576-128946576 on Build GRCh38
rs753651872 Chr.7: 128946617-128946617 on Build GRCh38
rs764747303 Chr.7: 128946545-128946553 on Build GRCh38
rs751194938 Chr.7: 128946455-128946455 on Build GRCh38
rs747040494 Chr.7: 128946545-128946545 on Build GRCh38
rs755529335 Chr.7: 128946608-128946608 on Build GRCh38
rs1041379928 Chr.7: 128946442-128946442 on Build GRCh38
rs759163802 Chr.7: 128946539-128946541 on Build GRCh38

View Full Product Details