Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 72464991-72465216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC6B
Entrez Gene ID
23233
Gene Name
exocyst complex component 6B
Gene Aliases
SEC15B, SEC15L2
UniGene
Hs.303454
Gene Chromosome Location
Chr.2: 72175984-72826042 on Build GRCh38
Transcripts
NM_001321729.1 NM_001321730.1 NM_001321731.1 NM_001321733.1 NM_001321734.1 NM_015189.2 NR_135773.1 NR_135774.1 XM_005264224.1 XM_011532711.2 XM_011532712.2 XM_017003641.1 XM_017003642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4315069 c.1940C>T p.A647V Chr.2: 72465200-72465200 on Build GRCh38
5736075 c.1954A>G p.T652A Chr.2: 72465186-72465186 on Build GRCh38
6467401 c.556C>T p.L186F Chr.2: 72465195-72465195 on Build GRCh38
5744527 c.1927G>C p.V643L Chr.2: 72465213-72465213 on Build GRCh38
4315070 c.551C>T p.A184V Chr.2: 72465200-72465200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965604388 Chr.2: 72465124-72465124 on Build GRCh38
rs372683988 Chr.2: 72465110-72465110 on Build GRCh38
rs746477892 Chr.2: 72465191-72465191 on Build GRCh38
rs397829667 Chr.2: 72465145-72465146 on Build GRCh38
rs1018128923 Chr.2: 72465117-72465117 on Build GRCh38
rs1032055165 Chr.2: 72465142-72465143 on Build GRCh38
rs867198508 Chr.2: 72465140-72465141 on Build GRCh38
rs35274285 Chr.2: 72465144-72465145 on Build GRCh38
rs888397190 Chr.2: 72465042-72465042 on Build GRCh38
rs572123657 Chr.2: 72465165-72465165 on Build GRCh38
rs765057443 Chr.2: 72465041-72465041 on Build GRCh38
rs56194993 Chr.2: 72465023-72465023 on Build GRCh38
rs374962579 Chr.2: 72465128-72465128 on Build GRCh38
rs869226440 Chr.2: 72465146-72465147 on Build GRCh38
rs5832083 Chr.2: 72465143-72465144 on Build GRCh38
rs747815795 Chr.2: 72465147-72465147 on Build GRCh38
rs978439565 Chr.2: 72465154-72465154 on Build GRCh38
rs749243251 Chr.2: 72465119-72465119 on Build GRCh38
rs1020463995 Chr.2: 72465130-72465130 on Build GRCh38
rs1006797461 Chr.2: 72465116-72465116 on Build GRCh38
rs897875656 Chr.2: 72465036-72465036 on Build GRCh38
rs368898383 Chr.2: 72465192-72465192 on Build GRCh38
rs994841256 Chr.2: 72465040-72465040 on Build GRCh38
rs772951078 Chr.2: 72465188-72465188 on Build GRCh38
rs751630500 Chr.2: 72465104-72465104 on Build GRCh38
rs560314747 Chr.2: 72465048-72465048 on Build GRCh38
rs759833934 Chr.2: 72465046-72465046 on Build GRCh38
rs758603001 Chr.2: 72465058-72465058 on Build GRCh38
rs757442879 Chr.2: 72465105-72465105 on Build GRCh38
rs778632495 Chr.2: 72465136-72465136 on Build GRCh38
rs866386569 Chr.2: 72465142-72465143 on Build GRCh38
rs201827100 Chr.2: 72465142-72465143 on Build GRCh38
rs764211609 Chr.2: 72465097-72465097 on Build GRCh38

View Full Product Details