Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 69005719-69005952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF1BP
Entrez Gene ID
26128
Gene Name
KIF1 binding protein
Gene Aliases
KBP, KIAA1279, TTC20
UniGene
Hs.279580
Gene Chromosome Location
Chr.10: 68988721-69016983 on Build GRCh38
Transcripts
NM_015634.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5901659 c.736C>T p.P246S Chr.10: 69005862-69005862 on Build GRCh38
5084071 c.767T>C p.L256S Chr.10: 69005893-69005893 on Build GRCh38
5662654 c.642A>G p.L214L Chr.10: 69005768-69005768 on Build GRCh38
4015340 c.703C>A p.L235I Chr.10: 69005829-69005829 on Build GRCh38
6365888 c.623C>G p.T208S Chr.10: 69005749-69005749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141052402 Chr.10: 69005854-69005854 on Build GRCh38
rs747600573 Chr.10: 69005790-69005790 on Build GRCh38
rs370670715 Chr.10: 69005782-69005782 on Build GRCh38
rs140826893 Chr.10: 69005859-69005859 on Build GRCh38
rs768162943 Chr.10: 69005770-69005770 on Build GRCh38
rs997199629 Chr.10: 69005840-69005840 on Build GRCh38
rs774544412 Chr.10: 69005819-69005819 on Build GRCh38
rs547582634 Chr.10: 69005844-69005844 on Build GRCh38
rs368916440 Chr.10: 69005795-69005795 on Build GRCh38
rs937189116 Chr.10: 69005882-69005882 on Build GRCh38
rs990183475 Chr.10: 69005807-69005807 on Build GRCh38
rs758386573 Chr.10: 69005846-69005846 on Build GRCh38
rs572485003 Chr.10: 69005904-69005904 on Build GRCh38
rs759865249 Chr.10: 69005823-69005823 on Build GRCh38
rs752833136 Chr.10: 69005835-69005835 on Build GRCh38
rs780651935 Chr.10: 69005881-69005881 on Build GRCh38
rs760604068 Chr.10: 69005836-69005836 on Build GRCh38
rs191873145 Chr.10: 69005814-69005814 on Build GRCh38
rs935472274 Chr.10: 69005773-69005773 on Build GRCh38
rs747701445 Chr.10: 69005895-69005895 on Build GRCh38
rs376375367 Chr.10: 69005831-69005831 on Build GRCh38
rs781257334 Chr.10: 69005805-69005805 on Build GRCh38
rs751308866 Chr.10: 69005855-69005855 on Build GRCh38
rs749110699 Chr.10: 69005907-69005907 on Build GRCh38
rs778904774 Chr.10: 69005903-69005903 on Build GRCh38
rs373081651 Chr.10: 69005810-69005810 on Build GRCh38
rs576197549 Chr.10: 69005804-69005804 on Build GRCh38
rs1016134527 Chr.10: 69005856-69005856 on Build GRCh38
rs866036954 Chr.10: 69005894-69005894 on Build GRCh38

View Full Product Details