Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 168038374-168038643 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF25
Entrez Gene ID
3834
Gene Name
kinesin family member 25
Gene Aliases
KNSL3
UniGene
Hs.150013
Gene Chromosome Location
Chr.6: 168002543-168045106 on Build GRCh38
Transcripts
NM_005355.3 NM_030615.2 XM_011535802.2 XM_011535803.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5351298 c.319C>A p.L107I Chr.6: 168038554-168038554 on Build GRCh38
1076070 c.344G>A p.S115N Chr.6: 168038579-168038579 on Build GRCh38
5404801 c.405C>T p.A135A Chr.6: 168038640-168038640 on Build GRCh38
6261307 c.351G>T p.K117N Chr.6: 168038586-168038586 on Build GRCh38
4803240 c.358G>T p.V120F Chr.6: 168038593-168038593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046586766 Chr.6: 168038609-168038609 on Build GRCh38
rs1044253326 Chr.6: 168038498-168038498 on Build GRCh38
rs558813439 Chr.6: 168038429-168038429 on Build GRCh38
rs376908446 Chr.6: 168038549-168038549 on Build GRCh38
rs773665254 Chr.6: 168038532-168038532 on Build GRCh38
rs144813171 Chr.6: 168038613-168038613 on Build GRCh38
rs759742921 Chr.6: 168038551-168038551 on Build GRCh38
rs553664581 Chr.6: 168038464-168038464 on Build GRCh38
rs575229772 Chr.6: 168038467-168038467 on Build GRCh38
rs1030887820 Chr.6: 168038435-168038435 on Build GRCh38
rs57457790 Chr.6: 168038439-168038439 on Build GRCh38
rs774085789 Chr.6: 168038570-168038570 on Build GRCh38
rs760827079 Chr.6: 168038536-168038536 on Build GRCh38
rs751450720 Chr.6: 168038571-168038571 on Build GRCh38
rs1030398985 Chr.6: 168038619-168038619 on Build GRCh38
rs916742812 Chr.6: 168038424-168038424 on Build GRCh38
rs556899448 Chr.6: 168038488-168038488 on Build GRCh38
rs543597555 Chr.6: 168038399-168038400 on Build GRCh38
rs537537453 Chr.6: 168038422-168038422 on Build GRCh38
rs781147084 Chr.6: 168038562-168038562 on Build GRCh38
rs998027534 Chr.6: 168038489-168038489 on Build GRCh38
rs149429830 Chr.6: 168038610-168038610 on Build GRCh38
rs772604978 Chr.6: 168038524-168038524 on Build GRCh38
rs757087872 Chr.6: 168038572-168038572 on Build GRCh38
rs1011424146 Chr.6: 168038479-168038479 on Build GRCh38
rs948204721 Chr.6: 168038426-168038426 on Build GRCh38
rs750340386 Chr.6: 168038587-168038587 on Build GRCh38
rs575156753 Chr.6: 168038548-168038548 on Build GRCh38
rs563878525 Chr.6: 168038556-168038556 on Build GRCh38
rs1018325944 Chr.6: 168038481-168038481 on Build GRCh38
rs941125398 Chr.6: 168038412-168038412 on Build GRCh38
rs753733695 Chr.6: 168038474-168038474 on Build GRCh38
rs553734095 Chr.6: 168038517-168038517 on Build GRCh38
rs535278908 Chr.6: 168038449-168038449 on Build GRCh38
rs778503468 Chr.6: 168038614-168038614 on Build GRCh38
rs374252442 Chr.6: 168038579-168038579 on Build GRCh38
rs541423845 Chr.6: 168038487-168038487 on Build GRCh38
rs373816022 Chr.6: 168038508-168038508 on Build GRCh38
rs755667428 Chr.6: 168038595-168038595 on Build GRCh38
rs867244872 Chr.6: 168038563-168038563 on Build GRCh38
rs34993752 Chr.6: 168038410-168038411 on Build GRCh38
rs368990891 Chr.6: 168038491-168038491 on Build GRCh38
rs1029569339 Chr.6: 168038501-168038501 on Build GRCh38
rs924204725 Chr.6: 168038451-168038451 on Build GRCh38
rs866396918 Chr.6: 168038580-168038580 on Build GRCh38
rs752419076 Chr.6: 168038523-168038523 on Build GRCh38
rs1006217580 Chr.6: 168038411-168038411 on Build GRCh38
rs200343577 Chr.6: 168038615-168038615 on Build GRCh38
rs750553787 Chr.6: 168038473-168038473 on Build GRCh38
rs955741158 Chr.6: 168038457-168038457 on Build GRCh38
rs139284594 Chr.6: 168038599-168038599 on Build GRCh38

View Full Product Details