Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 96390442-96390701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JRKL
Entrez Gene ID
8690
Gene Name
JRK-like
Gene Aliases
HHMJG
UniGene
Hs.105940
Gene Chromosome Location
Chr.11: 96389994-96393563 on Build GRCh38
Transcripts
NM_001261833.1 NM_003772.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7102881 c.49G>C p.D17H Chr.11: 96390698-96390698 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs999930072 Chr.11: 96390517-96390517 on Build GRCh38
rs763510933 Chr.11: 96390607-96390607 on Build GRCh38
rs761418962 Chr.11: 96390632-96390632 on Build GRCh38
rs750570328 Chr.11: 96390623-96390624 on Build GRCh38
rs758609812 Chr.11: 96390626-96390627 on Build GRCh38
rs1040677187 Chr.11: 96390579-96390579 on Build GRCh38
rs529186894 Chr.11: 96390474-96390474 on Build GRCh38
rs397763454 Chr.11: 96390571-96390572 on Build GRCh38
rs992465300 Chr.11: 96390596-96390596 on Build GRCh38
rs993350176 Chr.11: 96390666-96390666 on Build GRCh38
rs931127820 Chr.11: 96390554-96390554 on Build GRCh38
rs189538024 Chr.11: 96390573-96390573 on Build GRCh38
rs3842515 Chr.11: 96390570-96390571 on Build GRCh38
rs1010440357 Chr.11: 96390598-96390598 on Build GRCh38
rs564359024 Chr.11: 96390590-96390593 on Build GRCh38
rs941497777 Chr.11: 96390635-96390635 on Build GRCh38
rs554974375 Chr.11: 96390470-96390470 on Build GRCh38
rs201469823 Chr.11: 96390624-96390624 on Build GRCh38
rs371863374 Chr.11: 96390606-96390606 on Build GRCh38
rs547245059 Chr.11: 96390557-96390557 on Build GRCh38
rs371685864 Chr.11: 96390625-96390625 on Build GRCh38
rs1048719163 Chr.11: 96390564-96390564 on Build GRCh38
rs771569217 Chr.11: 96390608-96390608 on Build GRCh38
rs36049185 Chr.11: 96390498-96390499 on Build GRCh38
rs765071180 Chr.11: 96390649-96390649 on Build GRCh38
rs763795835 Chr.11: 96390627-96390627 on Build GRCh38
rs1034054126 Chr.11: 96390548-96390548 on Build GRCh38
rs753495805 Chr.11: 96390630-96390630 on Build GRCh38
rs199703021 Chr.11: 96390616-96390616 on Build GRCh38
rs918510201 Chr.11: 96390486-96390486 on Build GRCh38

View Full Product Details