Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37051855-37052084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD17
Entrez Gene ID
79734
Gene Name
potassium channel tetramerization domain containing 17
Gene Aliases
-
UniGene
Hs.517597
Gene Chromosome Location
Chr.22: 37051725-37063390 on Build GRCh38
Transcripts
NM_001282684.1 NM_001282685.1 NM_001282686.1 NM_024681.3 XM_005261741.3 XM_005261742.2 XM_005261743.2 XM_005261744.2 XM_011530374.2 XM_011530377.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5154402 c.186delG p.E64fs*26 Chr.22: 37051925-37051925 on Build GRCh38
5621080 c.148C>T p.L50F Chr.22: 37051908-37051908 on Build GRCh38
5154401 c.165delG p.E57fs*26 Chr.22: 37051925-37051925 on Build GRCh38
5621081 c.169C>T p.L57F Chr.22: 37051908-37051908 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1036310997 Chr.22: 37052030-37052030 on Build GRCh38
rs564421158 Chr.22: 37051979-37051979 on Build GRCh38
rs17852877 Chr.22: 37051890-37051890 on Build GRCh38
rs939954939 Chr.22: 37052012-37052012 on Build GRCh38
rs757897190 Chr.22: 37051981-37051981 on Build GRCh38
rs572333922 Chr.22: 37051904-37051904 on Build GRCh38
rs35975182 Chr.22: 37052056-37052057 on Build GRCh38
rs200219289 Chr.22: 37051980-37051980 on Build GRCh38
rs932685687 Chr.22: 37052051-37052051 on Build GRCh38
rs940731873 Chr.22: 37052023-37052023 on Build GRCh38
rs899135837 Chr.22: 37052037-37052037 on Build GRCh38
rs770268401 Chr.22: 37051907-37051907 on Build GRCh38
rs977042233 Chr.22: 37052040-37052040 on Build GRCh38
rs1049735057 Chr.22: 37052055-37052055 on Build GRCh38
rs749559654 Chr.22: 37051965-37051965 on Build GRCh38
rs927163013 Chr.22: 37051948-37051948 on Build GRCh38
rs777313125 Chr.22: 37051874-37051874 on Build GRCh38
rs1035369608 Chr.22: 37051898-37051898 on Build GRCh38
rs774675445 Chr.22: 37051978-37051978 on Build GRCh38
rs546208305 Chr.22: 37051973-37051973 on Build GRCh38
rs767686948 Chr.22: 37051991-37051991 on Build GRCh38
rs959934185 Chr.22: 37051919-37051919 on Build GRCh38
rs866747575 Chr.22: 37052000-37052000 on Build GRCh38
rs922632564 Chr.22: 37052049-37052049 on Build GRCh38
rs775911747 Chr.22: 37051956-37051956 on Build GRCh38
rs992858584 Chr.22: 37052033-37052033 on Build GRCh38
rs1004173250 Chr.22: 37051894-37051894 on Build GRCh38
rs746352966 Chr.22: 37051895-37051895 on Build GRCh38
rs550305743 Chr.22: 37052017-37052017 on Build GRCh38

View Full Product Details