Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62357604-62357875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF2A
Entrez Gene ID
3796
Gene Name
kinesin family member 2A
Gene Aliases
CDCBM3, HK2, KIF2
UniGene
Hs.558351
Gene Chromosome Location
Chr.5: 62306162-62387184 on Build GRCh38
Transcripts
NM_001098511.2 NM_001243952.1 NM_001243953.1 NM_004520.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5096881 c.591_592insTG p.R202fs*1 Chr.5: 62357708-62357709 on Build GRCh38
1069241 c.674G>A p.C225Y Chr.5: 62357710-62357710 on Build GRCh38
4773954 c.574-4C>T p.? Chr.5: 62357687-62357687 on Build GRCh38
5096880 c.672_673insTG p.R229fs*1 Chr.5: 62357708-62357709 on Build GRCh38
5067578 c.592_593delTG p.V201fs*7 Chr.5: 62357709-62357710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763787903 Chr.5: 62357680-62357680 on Build GRCh38
rs1025965339 Chr.5: 62357643-62357643 on Build GRCh38
rs762670010 Chr.5: 62357678-62357678 on Build GRCh38
rs768889692 Chr.5: 62357758-62357758 on Build GRCh38
rs1005965636 Chr.5: 62357765-62357765 on Build GRCh38
rs371403370 Chr.5: 62357786-62357786 on Build GRCh38
rs751148284 Chr.5: 62357681-62357681 on Build GRCh38
rs749562947 Chr.5: 62357755-62357755 on Build GRCh38
rs758637711 Chr.5: 62357763-62357764 on Build GRCh38
rs377203715 Chr.5: 62357778-62357778 on Build GRCh38
rs763845512 Chr.5: 62357788-62357788 on Build GRCh38
rs377475865 Chr.5: 62357751-62357751 on Build GRCh38
rs781206641 Chr.5: 62357705-62357705 on Build GRCh38
rs756657897 Chr.5: 62357696-62357696 on Build GRCh38
rs535959764 Chr.5: 62357841-62357841 on Build GRCh38
rs889761575 Chr.5: 62357633-62357633 on Build GRCh38
rs780333361 Chr.5: 62357790-62357790 on Build GRCh38
rs750831553 Chr.5: 62357709-62357710 on Build GRCh38
rs750318761 Chr.5: 62357727-62357727 on Build GRCh38
rs372490049 Chr.5: 62357776-62357776 on Build GRCh38
rs752433920 Chr.5: 62357677-62357677 on Build GRCh38
rs369031984 Chr.5: 62357839-62357839 on Build GRCh38
rs766644615 Chr.5: 62357676-62357676 on Build GRCh38
rs779793946 Chr.5: 62357754-62357754 on Build GRCh38
rs761159106 Chr.5: 62357663-62357663 on Build GRCh38
rs777154937 Chr.5: 62357780-62357780 on Build GRCh38
rs370317467 Chr.5: 62357760-62357760 on Build GRCh38
rs772031884 Chr.5: 62357774-62357774 on Build GRCh38
rs748272149 Chr.5: 62357767-62357767 on Build GRCh38
rs1035916945 Chr.5: 62357810-62357810 on Build GRCh38

View Full Product Details