Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53981910-53982159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG8
Entrez Gene ID
59283
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 8
Gene Aliases
-
UniGene
Hs.631595
Gene Chromosome Location
Chr.19: 53963036-53990215 on Build GRCh38
Transcripts
NM_031895.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4792339 c.552C>A p.N184K Chr.19: 53982123-53982123 on Build GRCh38
398997 c.559G>C p.E187Q Chr.19: 53982130-53982130 on Build GRCh38
6152044 c.570G>A p.P190P Chr.19: 53982141-53982141 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1026460250 Chr.19: 53981974-53981974 on Build GRCh38
rs951194154 Chr.19: 53982109-53982109 on Build GRCh38
rs774967866 Chr.19: 53982084-53982084 on Build GRCh38
rs370574892 Chr.19: 53982093-53982093 on Build GRCh38
rs775591051 Chr.19: 53982059-53982059 on Build GRCh38
rs745954972 Chr.19: 53982032-53982032 on Build GRCh38
rs374191792 Chr.19: 53982099-53982099 on Build GRCh38
rs770015412 Chr.19: 53982044-53982044 on Build GRCh38
rs767935825 Chr.19: 53982094-53982094 on Build GRCh38
rs1020616160 Chr.19: 53982038-53982038 on Build GRCh38
rs926561131 Chr.19: 53982020-53982031 on Build GRCh38
rs751475664 Chr.19: 53982130-53982130 on Build GRCh38
rs758180951 Chr.19: 53982126-53982126 on Build GRCh38
rs776169340 Chr.19: 53982096-53982096 on Build GRCh38
rs769210120 Chr.19: 53982075-53982075 on Build GRCh38
rs957165091 Chr.19: 53982026-53982026 on Build GRCh38
rs142463241 Chr.19: 53982128-53982128 on Build GRCh38
rs970423339 Chr.19: 53981993-53981993 on Build GRCh38
rs374083366 Chr.19: 53982068-53982068 on Build GRCh38
rs962325965 Chr.19: 53981939-53981939 on Build GRCh38
rs367756450 Chr.19: 53982123-53982123 on Build GRCh38
rs981741056 Chr.19: 53982020-53982025 on Build GRCh38
rs781381207 Chr.19: 53982031-53982031 on Build GRCh38
rs959272972 Chr.19: 53982020-53982020 on Build GRCh38
rs781021411 Chr.19: 53982141-53982141 on Build GRCh38
rs1034165688 Chr.19: 53982013-53982013 on Build GRCh38
rs986603776 Chr.19: 53982118-53982118 on Build GRCh38
rs757136397 Chr.19: 53982135-53982135 on Build GRCh38
rs749657719 Chr.19: 53982067-53982067 on Build GRCh38
rs12979869 Chr.19: 53982006-53982006 on Build GRCh38
rs764884536 Chr.19: 53982122-53982122 on Build GRCh38

View Full Product Details