Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42956155-42956396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AARSD1
Entrez Gene ID
80755
Gene Name
alanyl-tRNA synthetase domain containing 1
Gene Aliases
-
UniGene
Hs.317403
Gene Chromosome Location
Chr.17: 42950526-42964498 on Build GRCh38
Transcripts
NM_001261434.1 NM_001136042.2 NM_025267.3

Gene Symbol
PTGES3L-AARSD1
Entrez Gene ID
100885850
Gene Name
PTGES3L-AARSD1 readthrough
Gene Aliases
-
UniGene
Hs.317403
Gene Chromosome Location
Chr.17: 42950526-42980528 on Build GRCh38
Transcripts
NM_001261434.1 NM_001136042.2 NM_025267.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4066712 c.895C>T p.R299W Chr.17: 42956311-42956311 on Build GRCh38
4066711 c.1078C>T p.R360W Chr.17: 42956311-42956311 on Build GRCh38
6758474 c.610G>A p.V204I Chr.17: 42956257-42956257 on Build GRCh38
5952024 c.1073G>T p.S358I Chr.17: 42956316-42956316 on Build GRCh38
4460768 c.996C>T p.D332D Chr.17: 42956210-42956210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774488952 Chr.17: 42956321-42956321 on Build GRCh38
rs574315993 Chr.17: 42956311-42956311 on Build GRCh38
rs763286283 Chr.17: 42956207-42956207 on Build GRCh38
rs763733058 Chr.17: 42956208-42956208 on Build GRCh38
rs199585384 Chr.17: 42956281-42956281 on Build GRCh38
rs775802230 Chr.17: 42956200-42956200 on Build GRCh38
rs774195120 Chr.17: 42956229-42956229 on Build GRCh38
rs760619124 Chr.17: 42956351-42956351 on Build GRCh38
rs750402783 Chr.17: 42956256-42956256 on Build GRCh38
rs761574327 Chr.17: 42956247-42956247 on Build GRCh38
rs765897068 Chr.17: 42956258-42956258 on Build GRCh38
rs563637163 Chr.17: 42956357-42956357 on Build GRCh38
rs376824339 Chr.17: 42956373-42956373 on Build GRCh38
rs558257000 Chr.17: 42956195-42956195 on Build GRCh38
rs200793732 Chr.17: 42956372-42956372 on Build GRCh38
rs754268262 Chr.17: 42956194-42956194 on Build GRCh38
rs921335304 Chr.17: 42956223-42956223 on Build GRCh38
rs186386843 Chr.17: 42956369-42956369 on Build GRCh38
rs769933004 Chr.17: 42956310-42956310 on Build GRCh38
rs768912657 Chr.17: 42956320-42956320 on Build GRCh38
rs372546398 Chr.17: 42956264-42956264 on Build GRCh38
rs376428983 Chr.17: 42956190-42956190 on Build GRCh38
rs199745290 Chr.17: 42956277-42956277 on Build GRCh38
rs746395438 Chr.17: 42956303-42956303 on Build GRCh38
rs753332873 Chr.17: 42956356-42956356 on Build GRCh38
rs745909690 Chr.17: 42956191-42956191 on Build GRCh38
rs977254922 Chr.17: 42956205-42956205 on Build GRCh38
rs765516775 Chr.17: 42956200-42956200 on Build GRCh38
rs757599546 Chr.17: 42956286-42956286 on Build GRCh38
rs757396494 Chr.17: 42956197-42956198 on Build GRCh38
rs200220879 Chr.17: 42956249-42956249 on Build GRCh38
rs755582852 Chr.17: 42956257-42956257 on Build GRCh38
rs781748785 Chr.17: 42956300-42956300 on Build GRCh38
rs756687957 Chr.17: 42956374-42956374 on Build GRCh38
rs747398417 Chr.17: 42956179-42956179 on Build GRCh38
rs558991922 Chr.17: 42956278-42956278 on Build GRCh38
rs771454253 Chr.17: 42956185-42956185 on Build GRCh38
rs771796823 Chr.17: 42956338-42956338 on Build GRCh38
rs202044806 Chr.17: 42956355-42956355 on Build GRCh38
rs370562646 Chr.17: 42956347-42956347 on Build GRCh38
rs749398557 Chr.17: 42956312-42956312 on Build GRCh38
rs759015544 Chr.17: 42956360-42956360 on Build GRCh38

View Full Product Details