Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7096727-7096998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1RL
Entrez Gene ID
51279
Gene Name
complement C1r subcomponent like
Gene Aliases
C1RL1, C1RLP, C1r-LP, CLSPa
UniGene
Hs.631730
Gene Chromosome Location
Chr.12: 7093662-7109278 on Build GRCh38
Transcripts
NM_001297640.1 NM_001297642.1 NM_016546.3 XM_017019397.1 XM_017019398.1 XM_017019399.1 XM_017019400.1 XM_017019402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
943321 c.1009A>G p.S337G Chr.12: 7096846-7096846 on Build GRCh38
3359926 c.901G>T p.V301L Chr.12: 7096954-7096954 on Build GRCh38
3465080 c.1053G>T p.L351L Chr.12: 7096802-7096802 on Build GRCh38
6334928 c.885G>A p.K295K Chr.12: 7096970-7096970 on Build GRCh38
4458350 c.1082C>T p.P361L Chr.12: 7096773-7096773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756055186 Chr.12: 7096785-7096785 on Build GRCh38
rs200855206 Chr.12: 7096828-7096828 on Build GRCh38
rs763277062 Chr.12: 7096843-7096843 on Build GRCh38
rs768571629 Chr.12: 7096958-7096959 on Build GRCh38
rs35483961 Chr.12: 7096904-7096905 on Build GRCh38
rs757164011 Chr.12: 7096961-7096961 on Build GRCh38
rs765074890 Chr.12: 7096963-7096963 on Build GRCh38
rs759003415 Chr.12: 7096749-7096749 on Build GRCh38
rs781408421 Chr.12: 7096976-7096979 on Build GRCh38
rs759804213 Chr.12: 7096942-7096942 on Build GRCh38
rs376129178 Chr.12: 7096760-7096760 on Build GRCh38
rs746562099 Chr.12: 7096894-7096894 on Build GRCh38
rs111937138 Chr.12: 7096948-7096948 on Build GRCh38
rs916415091 Chr.12: 7096902-7096902 on Build GRCh38
rs747146443 Chr.12: 7096847-7096848 on Build GRCh38
rs761510122 Chr.12: 7096771-7096771 on Build GRCh38
rs143762434 Chr.12: 7096877-7096877 on Build GRCh38
rs777837442 Chr.12: 7096790-7096790 on Build GRCh38
rs773679493 Chr.12: 7096841-7096841 on Build GRCh38
rs767914609 Chr.12: 7096944-7096944 on Build GRCh38
rs778456831 Chr.12: 7096801-7096801 on Build GRCh38
rs148521163 Chr.12: 7096924-7096924 on Build GRCh38
rs752879493 Chr.12: 7096870-7096870 on Build GRCh38
rs758159531 Chr.12: 7096970-7096970 on Build GRCh38
rs201874025 Chr.12: 7096784-7096784 on Build GRCh38
rs113750373 Chr.12: 7096951-7096951 on Build GRCh38
rs201239060 Chr.12: 7096772-7096772 on Build GRCh38
rs775467984 Chr.12: 7096793-7096793 on Build GRCh38
rs139459588 Chr.12: 7096753-7096753 on Build GRCh38
rs551297918 Chr.12: 7096858-7096858 on Build GRCh38
rs779093421 Chr.12: 7096956-7096956 on Build GRCh38
rs770724397 Chr.12: 7096794-7096794 on Build GRCh38
rs771566519 Chr.12: 7096752-7096752 on Build GRCh38
rs149653325 Chr.12: 7096792-7096792 on Build GRCh38
rs771202397 Chr.12: 7096930-7096930 on Build GRCh38
rs760167194 Chr.12: 7096834-7096834 on Build GRCh38
rs770544200 Chr.12: 7096747-7096747 on Build GRCh38
rs755034239 Chr.12: 7096779-7096779 on Build GRCh38
rs778706829 Chr.12: 7096887-7096887 on Build GRCh38
rs1051368821 Chr.12: 7096840-7096840 on Build GRCh38
rs762993498 Chr.12: 7096933-7096933 on Build GRCh38
rs775820531 Chr.12: 7096947-7096947 on Build GRCh38
rs750845784 Chr.12: 7096767-7096767 on Build GRCh38
rs138272943 Chr.12: 7096893-7096893 on Build GRCh38
rs146147268 Chr.12: 7096835-7096835 on Build GRCh38
rs957812942 Chr.12: 7096797-7096797 on Build GRCh38
rs767506196 Chr.12: 7096865-7096865 on Build GRCh38
rs766716859 Chr.12: 7096844-7096844 on Build GRCh38
rs1006906956 Chr.12: 7096869-7096869 on Build GRCh38
rs745524395 Chr.12: 7096889-7096889 on Build GRCh38
rs150866407 Chr.12: 7096876-7096876 on Build GRCh38
rs199646073 Chr.12: 7096967-7096967 on Build GRCh38
rs756256292 Chr.12: 7096875-7096875 on Build GRCh38
rs753727531 Chr.12: 7096960-7096960 on Build GRCh38
rs771490370 Chr.12: 7096833-7096833 on Build GRCh38
rs758098356 Chr.12: 7096977-7096977 on Build GRCh38
rs956090887 Chr.12: 7096910-7096910 on Build GRCh38
rs967333591 Chr.12: 7096845-7096845 on Build GRCh38
rs774044185 Chr.12: 7096748-7096748 on Build GRCh38
rs751783230 Chr.12: 7096775-7096775 on Build GRCh38
rs773730603 Chr.12: 7096912-7096912 on Build GRCh38
rs1038542561 Chr.12: 7096820-7096820 on Build GRCh38
rs759736325 Chr.12: 7096861-7096861 on Build GRCh38
rs768132632 Chr.12: 7096911-7096911 on Build GRCh38

View Full Product Details