Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66515900-66516157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf46
Entrez Gene ID
254778
Gene Name
chromosome 8 open reading frame 46
Gene Aliases
-
UniGene
Hs.268869
Gene Chromosome Location
Chr.8: 66493256-66518524 on Build GRCh38
Transcripts
NM_152765.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4635622 c.562A>G p.T188A Chr.8: 66516014-66516014 on Build GRCh38
3650282 c.600C>T p.T200T Chr.8: 66516052-66516052 on Build GRCh38
4827976 c.466G>A p.E156K Chr.8: 66515918-66515918 on Build GRCh38
3650281 c.552G>A p.R184R Chr.8: 66516004-66516004 on Build GRCh38
4971556 c.540C>A p.P180P Chr.8: 66515992-66515992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768209161 Chr.8: 66515992-66515992 on Build GRCh38
rs747645866 Chr.8: 66516101-66516101 on Build GRCh38
rs758750642 Chr.8: 66516071-66516071 on Build GRCh38
rs753877182 Chr.8: 66516058-66516058 on Build GRCh38
rs777769380 Chr.8: 66515976-66515976 on Build GRCh38
rs746606235 Chr.8: 66516083-66516083 on Build GRCh38
rs776731663 Chr.8: 66516121-66516121 on Build GRCh38
rs16932952 Chr.8: 66515983-66515983 on Build GRCh38
rs765020183 Chr.8: 66515948-66515948 on Build GRCh38
rs373696105 Chr.8: 66516010-66516010 on Build GRCh38
rs750260511 Chr.8: 66515954-66515954 on Build GRCh38
rs1029201621 Chr.8: 66516040-66516040 on Build GRCh38
rs754362036 Chr.8: 66515979-66515979 on Build GRCh38
rs772551845 Chr.8: 66516068-66516068 on Build GRCh38
rs769645186 Chr.8: 66515999-66515999 on Build GRCh38
rs926480416 Chr.8: 66516135-66516135 on Build GRCh38
rs753006022 Chr.8: 66515965-66515965 on Build GRCh38
rs891972888 Chr.8: 66516031-66516031 on Build GRCh38
rs761766475 Chr.8: 66516125-66516125 on Build GRCh38
rs201264396 Chr.8: 66515985-66515985 on Build GRCh38
rs754707440 Chr.8: 66515986-66515987 on Build GRCh38
rs756193718 Chr.8: 66515972-66515972 on Build GRCh38
rs761756057 Chr.8: 66515996-66515996 on Build GRCh38
rs780707495 Chr.8: 66516096-66516096 on Build GRCh38
rs200736847 Chr.8: 66516081-66516081 on Build GRCh38
rs779190583 Chr.8: 66515984-66515984 on Build GRCh38
rs1044442404 Chr.8: 66515953-66515953 on Build GRCh38
rs752951022 Chr.8: 66516032-66516032 on Build GRCh38
rs766323693 Chr.8: 66516025-66516025 on Build GRCh38
rs776291425 Chr.8: 66516001-66516001 on Build GRCh38
rs17322752 Chr.8: 66516133-66516133 on Build GRCh38
rs974165916 Chr.8: 66516106-66516106 on Build GRCh38
rs750966355 Chr.8: 66516070-66516070 on Build GRCh38
rs147854635 Chr.8: 66515986-66515986 on Build GRCh38
rs139723976 Chr.8: 66516002-66516002 on Build GRCh38
rs776618843 Chr.8: 66515936-66515936 on Build GRCh38
rs372087831 Chr.8: 66516119-66516119 on Build GRCh38
rs758981792 Chr.8: 66515987-66515987 on Build GRCh38
rs74540254 Chr.8: 66516090-66516090 on Build GRCh38
rs761723738 Chr.8: 66515947-66515947 on Build GRCh38
rs953610034 Chr.8: 66516050-66516050 on Build GRCh38
rs376062010 Chr.8: 66516063-66516063 on Build GRCh38
rs773258721 Chr.8: 66516110-66516110 on Build GRCh38
rs561811742 Chr.8: 66515993-66515993 on Build GRCh38
rs370365687 Chr.8: 66515961-66515961 on Build GRCh38
rs575615728 Chr.8: 66516045-66516045 on Build GRCh38
rs760869033 Chr.8: 66516033-66516033 on Build GRCh38
rs770841536 Chr.8: 66516115-66516115 on Build GRCh38
rs767885860 Chr.8: 66515962-66515962 on Build GRCh38
rs544246461 Chr.8: 66516079-66516079 on Build GRCh38
rs749292321 Chr.8: 66516114-66516114 on Build GRCh38
rs199568191 Chr.8: 66516102-66516102 on Build GRCh38
rs764244776 Chr.8: 66516120-66516120 on Build GRCh38
rs547109778 Chr.8: 66516097-66516097 on Build GRCh38
rs757732888 Chr.8: 66516059-66516059 on Build GRCh38
rs774287394 Chr.8: 66516118-66516118 on Build GRCh38
rs983683600 Chr.8: 66516127-66516127 on Build GRCh38

View Full Product Details