Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120080806-120081051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICDL1
Entrez Gene ID
92558
Gene Name
BICD family like cargo adaptor 1
Gene Aliases
BICDR-1, BICDR1, CCDC64, H_267D11.1
UniGene
Hs.369763
Gene Chromosome Location
Chr.12: 119989806-120094495 on Build GRCh38
Transcripts
NM_207311.2 XM_005253991.4 XM_006719694.3 XM_006719696.3 XM_006719697.3 XM_011538999.2 XM_011539000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4526595 c.1404G>A p.Q468Q Chr.12: 120080982-120080982 on Build GRCh38
4039345 c.413G>A p.S138N Chr.12: 120080900-120080900 on Build GRCh38
7158364 c.449G>C p.R150P Chr.12: 120080936-120080936 on Build GRCh38
4203934 c.512G>A p.R171H Chr.12: 120080999-120080999 on Build GRCh38
4203932 c.428C>T p.A143V Chr.12: 120080915-120080915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765468277 Chr.12: 120080964-120080964 on Build GRCh38
rs765429423 Chr.12: 120080930-120080930 on Build GRCh38
rs752811109 Chr.12: 120081000-120081000 on Build GRCh38
rs770646746 Chr.12: 120081006-120081006 on Build GRCh38
rs1032012379 Chr.12: 120080947-120080947 on Build GRCh38
rs748797229 Chr.12: 120080910-120080910 on Build GRCh38
rs369172036 Chr.12: 120080997-120080997 on Build GRCh38
rs371725753 Chr.12: 120080847-120080847 on Build GRCh38
rs940774599 Chr.12: 120080880-120080880 on Build GRCh38
rs751645916 Chr.12: 120080998-120080998 on Build GRCh38
rs778496938 Chr.12: 120081011-120081011 on Build GRCh38
rs745384807 Chr.12: 120081013-120081013 on Build GRCh38
rs777077419 Chr.12: 120080943-120080943 on Build GRCh38
rs373363052 Chr.12: 120081001-120081001 on Build GRCh38
rs770394688 Chr.12: 120080917-120080917 on Build GRCh38
rs769478476 Chr.12: 120080883-120080883 on Build GRCh38
rs750660669 Chr.12: 120080967-120080967 on Build GRCh38
rs748210058 Chr.12: 120080991-120080991 on Build GRCh38
rs577466716 Chr.12: 120080960-120080960 on Build GRCh38
rs373275826 Chr.12: 120080936-120080936 on Build GRCh38
rs754854821 Chr.12: 120080999-120080999 on Build GRCh38
rs747954000 Chr.12: 120080882-120080882 on Build GRCh38
rs746901149 Chr.12: 120080860-120080860 on Build GRCh38
rs774018654 Chr.12: 120080926-120080926 on Build GRCh38
rs758448249 Chr.12: 120080846-120080846 on Build GRCh38
rs569155727 Chr.12: 120080866-120080866 on Build GRCh38
rs780900212 Chr.12: 120080877-120080877 on Build GRCh38
rs777352984 Chr.12: 120080900-120080900 on Build GRCh38
rs763260424 Chr.12: 120080971-120080971 on Build GRCh38
rs762546944 Chr.12: 120080877-120080879 on Build GRCh38
rs922196715 Chr.12: 120080838-120080838 on Build GRCh38
rs551362155 Chr.12: 120081003-120081003 on Build GRCh38
rs748841343 Chr.12: 120081004-120081004 on Build GRCh38

View Full Product Details