Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10189839-10190100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM52B
Entrez Gene ID
120939
Gene Name
transmembrane protein 52B
Gene Aliases
C12orf59
UniGene
Hs.226422
Gene Chromosome Location
Chr.12: 10170599-10191804 on Build GRCh38
Transcripts
NM_001079815.1 NM_153022.3 XM_006719029.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4545770 c.385G>A p.V129I Chr.12: 10190033-10190033 on Build GRCh38
4545771 c.445G>A p.V149I Chr.12: 10190033-10190033 on Build GRCh38
7394306 c.490G>T p.E164* Chr.12: 10190078-10190078 on Build GRCh38
934124 c.361G>A p.A121T Chr.12: 10190009-10190009 on Build GRCh38
4823868 c.285C>A p.I95I Chr.12: 10189933-10189933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755259107 Chr.12: 10190045-10190045 on Build GRCh38
rs779240379 Chr.12: 10189959-10189959 on Build GRCh38
rs542201515 Chr.12: 10190025-10190025 on Build GRCh38
rs756659721 Chr.12: 10189967-10189967 on Build GRCh38
rs775401474 Chr.12: 10189998-10189998 on Build GRCh38
rs904245539 Chr.12: 10189952-10189952 on Build GRCh38
rs202019424 Chr.12: 10190044-10190044 on Build GRCh38
rs534357086 Chr.12: 10189984-10189984 on Build GRCh38
rs201774853 Chr.12: 10189926-10189926 on Build GRCh38
rs183796625 Chr.12: 10190040-10190040 on Build GRCh38
rs775485374 Chr.12: 10189924-10189924 on Build GRCh38
rs769972724 Chr.12: 10189907-10189907 on Build GRCh38
rs756399303 Chr.12: 10189880-10189880 on Build GRCh38
rs745505195 Chr.12: 10189988-10189988 on Build GRCh38
rs756384863 Chr.12: 10189948-10189948 on Build GRCh38
rs375901455 Chr.12: 10189873-10189873 on Build GRCh38
rs377591052 Chr.12: 10189945-10189945 on Build GRCh38
rs377272370 Chr.12: 10189862-10189862 on Build GRCh38
rs761754619 Chr.12: 10189930-10189930 on Build GRCh38
rs200082927 Chr.12: 10189958-10189958 on Build GRCh38
rs777308706 Chr.12: 10189905-10189905 on Build GRCh38
rs772079638 Chr.12: 10189973-10189973 on Build GRCh38
rs757477047 Chr.12: 10189864-10189864 on Build GRCh38
rs754239124 Chr.12: 10189957-10189957 on Build GRCh38
rs146938916 Chr.12: 10190066-10190066 on Build GRCh38
rs199507184 Chr.12: 10189878-10189878 on Build GRCh38
rs769107289 Chr.12: 10189901-10189901 on Build GRCh38
rs780407207 Chr.12: 10189888-10189888 on Build GRCh38
rs113241696 Chr.12: 10189877-10189877 on Build GRCh38
rs780391062 Chr.12: 10189950-10189950 on Build GRCh38
rs759266505 Chr.12: 10189913-10189913 on Build GRCh38
rs748721726 Chr.12: 10189964-10189964 on Build GRCh38
rs774181721 Chr.12: 10189925-10189925 on Build GRCh38
rs746552314 Chr.12: 10189906-10189906 on Build GRCh38
rs975474771 Chr.12: 10190080-10190080 on Build GRCh38
rs781222288 Chr.12: 10189875-10189875 on Build GRCh38
rs370047660 Chr.12: 10189935-10189935 on Build GRCh38
rs370251414 Chr.12: 10190041-10190041 on Build GRCh38
rs769639754 Chr.12: 10189995-10189995 on Build GRCh38
rs190704935 Chr.12: 10189999-10189999 on Build GRCh38
rs780847560 Chr.12: 10190072-10190072 on Build GRCh38
rs550537150 Chr.12: 10190035-10190035 on Build GRCh38
rs200896954 Chr.12: 10189946-10189946 on Build GRCh38
rs774085376 Chr.12: 10190020-10190020 on Build GRCh38
rs141227819 Chr.12: 10190049-10190049 on Build GRCh38
rs992737168 Chr.12: 10190009-10190009 on Build GRCh38
rs761823397 Chr.12: 10190024-10190024 on Build GRCh38
rs753092205 Chr.12: 10190064-10190064 on Build GRCh38
rs77905236 Chr.12: 10190032-10190032 on Build GRCh38
rs1034145258 Chr.12: 10189970-10189970 on Build GRCh38
rs569430362 Chr.12: 10189874-10189874 on Build GRCh38
rs769655936 Chr.12: 10189918-10189918 on Build GRCh38

View Full Product Details