Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180911219-180911492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL8
Entrez Gene ID
79908
Gene Name
butyrophilin like 8
Gene Aliases
BTN9.2
UniGene
Hs.189109
Gene Chromosome Location
Chr.5: 180899077-180952166 on Build GRCh38
Transcripts
NM_001040462.2 NM_001159707.1 NM_001159708.1 NM_001159709.1 NM_001159710.1 NM_024850.2 XM_011534649.1 XM_011534650.2 XM_011534651.2 XM_011534652.2 XM_011534653.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
737441 c.465G>T p.Q155H Chr.5: 180911406-180911406 on Build GRCh38
1696848 c.475T>C p.W159R Chr.5: 180911416-180911416 on Build GRCh38
6535845 c.491C>G p.T164R Chr.5: 180911432-180911432 on Build GRCh38
6221165 c.437T>C p.V146A Chr.5: 180911378-180911378 on Build GRCh38
6170852 c.494C>A p.A165E Chr.5: 180911435-180911435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993846730 Chr.5: 180911253-180911253 on Build GRCh38
rs2276995 Chr.5: 180911368-180911368 on Build GRCh38
rs749371133 Chr.5: 180911289-180911289 on Build GRCh38
rs937173927 Chr.5: 180911442-180911442 on Build GRCh38
rs754388874 Chr.5: 180911436-180911436 on Build GRCh38
rs746794728 Chr.5: 180911398-180911398 on Build GRCh38
rs766111997 Chr.5: 180911425-180911425 on Build GRCh38
rs368011336 Chr.5: 180911318-180911318 on Build GRCh38
rs768029042 Chr.5: 180911356-180911356 on Build GRCh38
rs776063430 Chr.5: 180911336-180911336 on Build GRCh38
rs756340453 Chr.5: 180911350-180911350 on Build GRCh38
rs976064102 Chr.5: 180911341-180911341 on Build GRCh38
rs386695827 Chr.5: 180911368-180911370 on Build GRCh38
rs375136243 Chr.5: 180911418-180911418 on Build GRCh38
rs148878394 Chr.5: 180911370-180911370 on Build GRCh38
rs146970792 Chr.5: 180911426-180911426 on Build GRCh38
rs754776707 Chr.5: 180911380-180911380 on Build GRCh38
rs761599158 Chr.5: 180911414-180911414 on Build GRCh38
rs778608424 Chr.5: 180911417-180911417 on Build GRCh38
rs952616016 Chr.5: 180911298-180911298 on Build GRCh38
rs532508917 Chr.5: 180911270-180911270 on Build GRCh38
rs758677132 Chr.5: 180911443-180911443 on Build GRCh38
rs746118211 Chr.5: 180911375-180911375 on Build GRCh38
rs372363727 Chr.5: 180911461-180911461 on Build GRCh38
rs372045915 Chr.5: 180911374-180911374 on Build GRCh38
rs749688648 Chr.5: 180911369-180911369 on Build GRCh38
rs774635510 Chr.5: 180911296-180911296 on Build GRCh38
rs199737855 Chr.5: 180911444-180911444 on Build GRCh38
rs765775195 Chr.5: 180911316-180911317 on Build GRCh38
rs750107545 Chr.5: 180911344-180911344 on Build GRCh38
rs534458966 Chr.5: 180911412-180911412 on Build GRCh38
rs1018602202 Chr.5: 180911316-180911316 on Build GRCh38
rs369838126 Chr.5: 180911377-180911377 on Build GRCh38
rs762739488 Chr.5: 180911421-180911421 on Build GRCh38
rs138408550 Chr.5: 180911468-180911468 on Build GRCh38
rs370167264 Chr.5: 180911435-180911435 on Build GRCh38
rs763995785 Chr.5: 180911319-180911319 on Build GRCh38
rs1041670938 Chr.5: 180911251-180911251 on Build GRCh38
rs764586330 Chr.5: 180911343-180911343 on Build GRCh38
rs779088856 Chr.5: 180911438-180911438 on Build GRCh38
rs775312208 Chr.5: 180911381-180911381 on Build GRCh38
rs2276994 Chr.5: 180911300-180911300 on Build GRCh38
rs761383953 Chr.5: 180911342-180911342 on Build GRCh38
rs775223793 Chr.5: 180911314-180911314 on Build GRCh38
rs761121379 Chr.5: 180911428-180911428 on Build GRCh38
rs758077353 Chr.5: 180911346-180911346 on Build GRCh38
rs148011374 Chr.5: 180911411-180911411 on Build GRCh38
rs756739021 Chr.5: 180911367-180911367 on Build GRCh38
rs200714281 Chr.5: 180911440-180911440 on Build GRCh38
rs753083807 Chr.5: 180911365-180911365 on Build GRCh38

View Full Product Details