Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179122555-179122816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS2
Entrez Gene ID
9509
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 2
Gene Aliases
ADAM-TS2, ADAMTS-2, ADAMTS-3, NPI, PC I-NP, PCI-NP, PCINP, PCPNI, PNPI
UniGene
Hs.23871
Gene Chromosome Location
Chr.5: 179110851-179345430 on Build GRCh38
Transcripts
NM_014244.4 XM_017010063.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4889948 c.3000G>T p.V1000V Chr.5: 179122732-179122732 on Build GRCh38
4763689 c.3088+1G>A p.? Chr.5: 179122643-179122643 on Build GRCh38
5500388 c.3077G>A p.G1026D Chr.5: 179122655-179122655 on Build GRCh38
4485862 c.2985C>T p.T995T Chr.5: 179122747-179122747 on Build GRCh38
4763690 c.3035G>A p.C1012Y Chr.5: 179122697-179122697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148344708 Chr.5: 179122656-179122656 on Build GRCh38
rs886060491 Chr.5: 179122654-179122654 on Build GRCh38
rs757846382 Chr.5: 179122632-179122632 on Build GRCh38
rs376454904 Chr.5: 179122709-179122709 on Build GRCh38
rs773933916 Chr.5: 179122712-179122712 on Build GRCh38
rs755241503 Chr.5: 179122748-179122748 on Build GRCh38
rs952120390 Chr.5: 179122606-179122606 on Build GRCh38
rs564326576 Chr.5: 179122585-179122585 on Build GRCh38
rs531483121 Chr.5: 179122587-179122587 on Build GRCh38
rs773184209 Chr.5: 179122773-179122773 on Build GRCh38
rs966437723 Chr.5: 179122644-179122644 on Build GRCh38
rs371013989 Chr.5: 179122694-179122694 on Build GRCh38
rs567951590 Chr.5: 179122592-179122592 on Build GRCh38
rs769744215 Chr.5: 179122700-179122700 on Build GRCh38
rs761101596 Chr.5: 179122713-179122713 on Build GRCh38
rs748783596 Chr.5: 179122780-179122780 on Build GRCh38
rs571054777 Chr.5: 179122720-179122720 on Build GRCh38
rs776489447 Chr.5: 179122687-179122687 on Build GRCh38
rs747970179 Chr.5: 179122753-179122753 on Build GRCh38
rs6863024 Chr.5: 179122789-179122789 on Build GRCh38
rs750505827 Chr.5: 179122725-179122725 on Build GRCh38
rs994072278 Chr.5: 179122591-179122591 on Build GRCh38
rs747134449 Chr.5: 179122745-179122745 on Build GRCh38
rs374871196 Chr.5: 179122619-179122619 on Build GRCh38
rs759056190 Chr.5: 179122786-179122786 on Build GRCh38
rs61754844 Chr.5: 179122672-179122672 on Build GRCh38
rs781386762 Chr.5: 179122659-179122659 on Build GRCh38
rs757793067 Chr.5: 179122719-179122719 on Build GRCh38
rs769689633 Chr.5: 179122762-179122762 on Build GRCh38
rs772670607 Chr.5: 179122703-179122703 on Build GRCh38
rs898087922 Chr.5: 179122732-179122732 on Build GRCh38
rs758462431 Chr.5: 179122731-179122731 on Build GRCh38
rs754044289 Chr.5: 179122727-179122727 on Build GRCh38
rs762314986 Chr.5: 179122798-179122798 on Build GRCh38
rs750487311 Chr.5: 179122686-179122686 on Build GRCh38
rs766033559 Chr.5: 179122705-179122705 on Build GRCh38
rs142429109 Chr.5: 179122752-179122752 on Build GRCh38
rs780314895 Chr.5: 179122738-179122738 on Build GRCh38
rs780184702 Chr.5: 179122684-179122684 on Build GRCh38
rs1027799593 Chr.5: 179122604-179122604 on Build GRCh38
rs79330641 Chr.5: 179122718-179122718 on Build GRCh38
rs770419188 Chr.5: 179122784-179122784 on Build GRCh38
rs1027851899 Chr.5: 179122772-179122772 on Build GRCh38
rs546990509 Chr.5: 179122653-179122653 on Build GRCh38
rs746802388 Chr.5: 179122685-179122685 on Build GRCh38
rs910578729 Chr.5: 179122629-179122629 on Build GRCh38
rs2303641 Chr.5: 179122790-179122790 on Build GRCh38
rs368690576 Chr.5: 179122704-179122704 on Build GRCh38
rs376403895 Chr.5: 179122623-179122623 on Build GRCh38
rs773920252 Chr.5: 179122785-179122785 on Build GRCh38
rs550082602 Chr.5: 179122589-179122589 on Build GRCh38
rs150535792 Chr.5: 179122714-179122714 on Build GRCh38

View Full Product Details