Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95924093-95924362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf131
Entrez Gene ID
100127889
Gene Name
chromosome 10 open reading frame 131
Gene Aliases
bA690P14.3
UniGene
Hs.719076
Gene Chromosome Location
Chr.10: 95907965-95938658 on Build GRCh38
Transcripts
NM_001130446.2 NR_038444.1

Gene Symbol
ENTPD1-AS1
Entrez Gene ID
728558
Gene Name
ENTPD1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.538374
Gene Chromosome Location
Chr.10: 95753206-96090235 on Build GRCh38
Transcripts
NM_001130446.2 NR_038444.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4830426 c.82C>G p.Q28E Chr.10: 95924334-95924334 on Build GRCh38
466133 c.82C>A p.Q28K Chr.10: 95924334-95924334 on Build GRCh38
4830427 c.82C>G p.Q28E Chr.10: 95924334-95924334 on Build GRCh38
4830428 c.94C>G p.Q32E Chr.10: 95924334-95924334 on Build GRCh38
1135075 c.94C>A p.Q32K Chr.10: 95924334-95924334 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748510619 Chr.10: 95924221-95924221 on Build GRCh38
rs752649660 Chr.10: 95924275-95924275 on Build GRCh38
rs1023117666 Chr.10: 95924224-95924224 on Build GRCh38
rs774869762 Chr.10: 95924181-95924181 on Build GRCh38
rs11188521 Chr.10: 95924157-95924157 on Build GRCh38
rs74503287 Chr.10: 95924150-95924150 on Build GRCh38
rs991199891 Chr.10: 95924190-95924190 on Build GRCh38
rs1002690749 Chr.10: 95924231-95924231 on Build GRCh38
rs182817770 Chr.10: 95924189-95924189 on Build GRCh38
rs968047796 Chr.10: 95924311-95924311 on Build GRCh38
rs957016639 Chr.10: 95924153-95924153 on Build GRCh38
rs764040294 Chr.10: 95924256-95924258 on Build GRCh38
rs200081023 Chr.10: 95924238-95924238 on Build GRCh38
rs561158812 Chr.10: 95924250-95924250 on Build GRCh38
rs1044160825 Chr.10: 95924193-95924193 on Build GRCh38
rs529967318 Chr.10: 95924297-95924297 on Build GRCh38
rs1020928169 Chr.10: 95924282-95924282 on Build GRCh38
rs374333694 Chr.10: 95924279-95924279 on Build GRCh38
rs891707855 Chr.10: 95924270-95924270 on Build GRCh38

View Full Product Details