Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58508834-58509095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOX2
Entrez Gene ID
8309
Gene Name
acyl-CoA oxidase 2
Gene Aliases
BCOX, BRCACOX, BRCOX, THCCox
UniGene
Hs.444959
Gene Chromosome Location
Chr.3: 58505136-58537202 on Build GRCh38
Transcripts
NM_003500.3 XM_005265505.1 XM_006713340.3 XM_017007302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6165299 c.1892A>G p.D631G Chr.3: 58508984-58508984 on Build GRCh38
6365372 c.1975A>T p.N659Y Chr.3: 58508901-58508901 on Build GRCh38
2782772 c.1942C>T p.R648C Chr.3: 58508934-58508934 on Build GRCh38
7084447 c.1939G>A p.E647K Chr.3: 58508937-58508937 on Build GRCh38
1047809 c.1938C>T p.Y646Y Chr.3: 58508938-58508938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756752270 Chr.3: 58508934-58508934 on Build GRCh38
rs778657401 Chr.3: 58508943-58508943 on Build GRCh38
rs542643101 Chr.3: 58508981-58508981 on Build GRCh38
rs917523468 Chr.3: 58508918-58508918 on Build GRCh38
rs1049479762 Chr.3: 58508878-58508878 on Build GRCh38
rs562396036 Chr.3: 58509062-58509062 on Build GRCh38
rs373685563 Chr.3: 58509065-58509065 on Build GRCh38
rs749223348 Chr.3: 58508867-58508867 on Build GRCh38
rs747212979 Chr.3: 58508955-58508955 on Build GRCh38
rs763344658 Chr.3: 58508996-58508996 on Build GRCh38
rs760869269 Chr.3: 58508888-58508888 on Build GRCh38
rs755771879 Chr.3: 58509040-58509041 on Build GRCh38
rs747884841 Chr.3: 58508883-58508883 on Build GRCh38
rs371093489 Chr.3: 58508953-58508953 on Build GRCh38
rs187194712 Chr.3: 58508886-58508886 on Build GRCh38
rs756621386 Chr.3: 58509004-58509004 on Build GRCh38
rs771014585 Chr.3: 58508890-58508890 on Build GRCh38
rs755667073 Chr.3: 58508935-58508935 on Build GRCh38
rs373278690 Chr.3: 58508868-58508868 on Build GRCh38
rs776770801 Chr.3: 58508967-58508967 on Build GRCh38
rs367780739 Chr.3: 58509059-58509059 on Build GRCh38
rs777054443 Chr.3: 58508911-58508911 on Build GRCh38
rs753783991 Chr.3: 58508938-58508938 on Build GRCh38
rs753401525 Chr.3: 58509040-58509040 on Build GRCh38
rs950300380 Chr.3: 58508929-58508929 on Build GRCh38
rs992011493 Chr.3: 58509066-58509066 on Build GRCh38
rs770977337 Chr.3: 58508963-58508963 on Build GRCh38
rs984566222 Chr.3: 58508859-58508859 on Build GRCh38
rs753226045 Chr.3: 58508900-58508903 on Build GRCh38
rs376522814 Chr.3: 58508874-58508874 on Build GRCh38
rs199808000 Chr.3: 58508937-58508937 on Build GRCh38
rs767185556 Chr.3: 58508924-58508924 on Build GRCh38
rs765251274 Chr.3: 58509049-58509049 on Build GRCh38
rs573721661 Chr.3: 58508944-58508944 on Build GRCh38
rs147680655 Chr.3: 58508916-58508916 on Build GRCh38
rs764417199 Chr.3: 58509001-58509001 on Build GRCh38
rs745872186 Chr.3: 58508974-58508974 on Build GRCh38
rs775931712 Chr.3: 58508986-58508986 on Build GRCh38
rs754945523 Chr.3: 58508942-58508942 on Build GRCh38
rs778762236 Chr.3: 58508880-58508880 on Build GRCh38
rs760369877 Chr.3: 58509038-58509038 on Build GRCh38
rs763249170 Chr.3: 58508921-58508921 on Build GRCh38
rs753068889 Chr.3: 58508919-58508919 on Build GRCh38
rs992205976 Chr.3: 58508899-58508899 on Build GRCh38
rs754397355 Chr.3: 58509044-58509044 on Build GRCh38
rs777345885 Chr.3: 58509012-58509012 on Build GRCh38
rs777700413 Chr.3: 58509063-58509063 on Build GRCh38
rs200638638 Chr.3: 58509039-58509039 on Build GRCh38
rs772905945 Chr.3: 58509037-58509037 on Build GRCh38
rs370979089 Chr.3: 58508885-58508885 on Build GRCh38
rs758228481 Chr.3: 58508948-58508948 on Build GRCh38
rs760077878 Chr.3: 58508914-58508914 on Build GRCh38

View Full Product Details