Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34883172-34883433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APIP
Entrez Gene ID
51074
Gene Name
APAF1 interacting protein
Gene Aliases
APIP2, CGI-29, CGI29, MMRP19, hAPIP
UniGene
Hs.447794
Gene Chromosome Location
Chr.11: 34882295-34916411 on Build GRCh38
Transcripts
NM_015957.3 XM_011520154.2 XM_017017875.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6360143 c.580C>T p.R194C Chr.11: 34883386-34883386 on Build GRCh38
4787250 c.648G>T p.V216V Chr.11: 34883369-34883369 on Build GRCh38
4502763 c.621C>A p.A207A Chr.11: 34883345-34883345 on Build GRCh38
1638825 c.618G>A p.K206K Chr.11: 34883348-34883348 on Build GRCh38
6132440 c.621A>T p.V207V Chr.11: 34883396-34883396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770989795 Chr.11: 34883381-34883381 on Build GRCh38
rs761971765 Chr.11: 34883327-34883327 on Build GRCh38
rs749540515 Chr.11: 34883228-34883228 on Build GRCh38
rs768975119 Chr.11: 34883364-34883364 on Build GRCh38
rs146983826 Chr.11: 34883296-34883296 on Build GRCh38
rs769894777 Chr.11: 34883375-34883375 on Build GRCh38
rs745603170 Chr.11: 34883367-34883367 on Build GRCh38
rs373309162 Chr.11: 34883332-34883332 on Build GRCh38
rs773813457 Chr.11: 34883396-34883397 on Build GRCh38
rs780136237 Chr.11: 34883372-34883372 on Build GRCh38
rs753257197 Chr.11: 34883356-34883356 on Build GRCh38
rs745692899 Chr.11: 34883304-34883304 on Build GRCh38
rs542086947 Chr.11: 34883278-34883278 on Build GRCh38
rs1050257201 Chr.11: 34883246-34883246 on Build GRCh38
rs201621404 Chr.11: 34883403-34883403 on Build GRCh38
rs866295365 Chr.11: 34883366-34883367 on Build GRCh38
rs930068374 Chr.11: 34883237-34883237 on Build GRCh38
rs934202058 Chr.11: 34883243-34883243 on Build GRCh38
rs763132696 Chr.11: 34883337-34883337 on Build GRCh38
rs564123868 Chr.11: 34883362-34883362 on Build GRCh38
rs765301669 Chr.11: 34883331-34883331 on Build GRCh38
rs200074052 Chr.11: 34883359-34883359 on Build GRCh38
rs773115995 Chr.11: 34883377-34883377 on Build GRCh38
rs1014060494 Chr.11: 34883370-34883370 on Build GRCh38
rs768499452 Chr.11: 34883367-34883368 on Build GRCh38
rs184470334 Chr.11: 34883248-34883248 on Build GRCh38
rs138117165 Chr.11: 34883343-34883343 on Build GRCh38
rs190618941 Chr.11: 34883353-34883353 on Build GRCh38
rs552847542 Chr.11: 34883200-34883200 on Build GRCh38
rs1055053545 Chr.11: 34883315-34883315 on Build GRCh38
rs746983925 Chr.11: 34883374-34883374 on Build GRCh38
rs775583617 Chr.11: 34883322-34883322 on Build GRCh38
rs766645188 Chr.11: 34883338-34883338 on Build GRCh38
rs755631949 Chr.11: 34883292-34883292 on Build GRCh38
rs778715503 Chr.11: 34883365-34883365 on Build GRCh38
rs112184665 Chr.11: 34883311-34883311 on Build GRCh38
rs759877175 Chr.11: 34883386-34883386 on Build GRCh38
rs193139398 Chr.11: 34883385-34883385 on Build GRCh38
rs751880596 Chr.11: 34883339-34883339 on Build GRCh38
rs777465058 Chr.11: 34883295-34883295 on Build GRCh38
rs533430185 Chr.11: 34883380-34883380 on Build GRCh38

View Full Product Details