Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102731827-102732062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC168
Entrez Gene ID
643677
Gene Name
coiled-coil domain containing 168
Gene Aliases
C13orf40
UniGene
Hs.368382
Gene Chromosome Location
Chr.13: 102729367-102759072 on Build GRCh38
Transcripts
NM_001146197.1 XM_011521106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1365264 c.4905C>T p.I1635I Chr.13: 102731905-102731905 on Build GRCh38
5670297 c.4981C>G p.P1661A Chr.13: 102731829-102731829 on Build GRCh38
3417331 c.4820A>G p.K1607R Chr.13: 102731990-102731990 on Build GRCh38
1365265 c.4821delA p.K1607fs*3 Chr.13: 102731989-102731989 on Build GRCh38
4650137 c.4838C>T p.T1613I Chr.13: 102731972-102731972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774117233 Chr.13: 102731980-102731980 on Build GRCh38
rs536877767 Chr.13: 102731879-102731879 on Build GRCh38
rs960283297 Chr.13: 102731869-102731869 on Build GRCh38
rs555275413 Chr.13: 102731961-102731961 on Build GRCh38
rs1003516335 Chr.13: 102732013-102732013 on Build GRCh38
rs888815149 Chr.13: 102731989-102731989 on Build GRCh38
rs1011325089 Chr.13: 102732028-102732028 on Build GRCh38
rs745818204 Chr.13: 102731928-102731928 on Build GRCh38
rs566264361 Chr.13: 102731868-102731868 on Build GRCh38
rs752940990 Chr.13: 102731866-102731866 on Build GRCh38
rs748979990 Chr.13: 102732036-102732036 on Build GRCh38
rs537011276 Chr.13: 102732018-102732018 on Build GRCh38
rs186542518 Chr.13: 102731964-102731964 on Build GRCh38
rs781160708 Chr.13: 102731908-102731908 on Build GRCh38
rs768405500 Chr.13: 102731972-102731972 on Build GRCh38
rs775384324 Chr.13: 102731963-102731963 on Build GRCh38
rs908712754 Chr.13: 102731906-102731906 on Build GRCh38
rs952785392 Chr.13: 102731904-102731904 on Build GRCh38
rs894273065 Chr.13: 102731889-102731889 on Build GRCh38
rs757320027 Chr.13: 102731905-102731905 on Build GRCh38
rs758682044 Chr.13: 102731876-102731876 on Build GRCh38
rs765205143 Chr.13: 102731912-102731912 on Build GRCh38
rs761722559 Chr.13: 102731880-102731880 on Build GRCh38

View Full Product Details