Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 81330335-81330608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT12
Entrez Gene ID
134526
Gene Name
acyl-CoA thioesterase 12
Gene Aliases
CACH-1, Cach, STARD15, THEAL
UniGene
Hs.591756
Gene Chromosome Location
Chr.5: 81309045-81394174 on Build GRCh38
Transcripts
NM_130767.2 XM_006714531.2 XM_017009045.1 XM_017009046.1 XM_017009047.1 XM_017009048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
308819 c.1560C>A p.Y520* Chr.5: 81330502-81330502 on Build GRCh38
4658861 c.1568G>T p.G523V Chr.5: 81330494-81330494 on Build GRCh38
4649744 c.1610C>G p.A537G Chr.5: 81330452-81330452 on Build GRCh38
461941 c.1573C>G p.L525V Chr.5: 81330489-81330489 on Build GRCh38
4916524 c.1663T>G p.F555V Chr.5: 81330399-81330399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778446106 Chr.5: 81330417-81330417 on Build GRCh38
rs754595354 Chr.5: 81330414-81330414 on Build GRCh38
rs766326711 Chr.5: 81330543-81330543 on Build GRCh38
rs1045405347 Chr.5: 81330501-81330501 on Build GRCh38
rs577790866 Chr.5: 81330448-81330448 on Build GRCh38
rs763221562 Chr.5: 81330530-81330530 on Build GRCh38
rs938139797 Chr.5: 81330444-81330444 on Build GRCh38
rs750383961 Chr.5: 81330470-81330470 on Build GRCh38
rs767383461 Chr.5: 81330391-81330391 on Build GRCh38
rs540665090 Chr.5: 81330443-81330443 on Build GRCh38
rs773600898 Chr.5: 81330430-81330430 on Build GRCh38
rs777328911 Chr.5: 81330512-81330512 on Build GRCh38
rs753609339 Chr.5: 81330500-81330500 on Build GRCh38
rs915104370 Chr.5: 81330496-81330496 on Build GRCh38
rs935456105 Chr.5: 81330465-81330465 on Build GRCh38
rs73768070 Chr.5: 81330560-81330560 on Build GRCh38
rs750307867 Chr.5: 81330392-81330392 on Build GRCh38
rs767778920 Chr.5: 81330527-81330527 on Build GRCh38
rs771483296 Chr.5: 81330452-81330452 on Build GRCh38
rs746264273 Chr.5: 81330526-81330526 on Build GRCh38
rs377706612 Chr.5: 81330540-81330540 on Build GRCh38
rs200890414 Chr.5: 81330499-81330499 on Build GRCh38
rs757862981 Chr.5: 81330511-81330511 on Build GRCh38
rs753737160 Chr.5: 81330548-81330548 on Build GRCh38
rs775737799 Chr.5: 81330461-81330461 on Build GRCh38
rs759331443 Chr.5: 81330557-81330557 on Build GRCh38
rs759699810 Chr.5: 81330471-81330472 on Build GRCh38
rs376149948 Chr.5: 81330502-81330502 on Build GRCh38
rs768684275 Chr.5: 81330531-81330531 on Build GRCh38
rs560879781 Chr.5: 81330446-81330446 on Build GRCh38
rs766461929 Chr.5: 81330405-81330405 on Build GRCh38
rs765574953 Chr.5: 81330456-81330456 on Build GRCh38
rs760600444 Chr.5: 81330492-81330492 on Build GRCh38
rs374522724 Chr.5: 81330536-81330536 on Build GRCh38
rs752330587 Chr.5: 81330510-81330510 on Build GRCh38
rs775587113 Chr.5: 81330529-81330529 on Build GRCh38
rs781743795 Chr.5: 81330519-81330519 on Build GRCh38
rs374342286 Chr.5: 81330533-81330533 on Build GRCh38
rs962362940 Chr.5: 81330566-81330566 on Build GRCh38
rs778568996 Chr.5: 81330509-81330509 on Build GRCh38
rs747635244 Chr.5: 81330418-81330418 on Build GRCh38
rs759557236 Chr.5: 81330406-81330406 on Build GRCh38
rs760655686 Chr.5: 81330538-81330538 on Build GRCh38
rs536125060 Chr.5: 81330467-81330467 on Build GRCh38
rs771572593 Chr.5: 81330420-81330420 on Build GRCh38
rs532812999 Chr.5: 81330453-81330453 on Build GRCh38
rs752799749 Chr.5: 81330375-81330375 on Build GRCh38
rs779809926 Chr.5: 81330401-81330401 on Build GRCh38
rs142957507 Chr.5: 81330455-81330455 on Build GRCh38
rs148630854 Chr.5: 81330515-81330515 on Build GRCh38
rs749006398 Chr.5: 81330407-81330407 on Build GRCh38
rs758450906 Chr.5: 81330376-81330376 on Build GRCh38
rs762945959 Chr.5: 81330466-81330466 on Build GRCh38
rs1036063682 Chr.5: 81330554-81330554 on Build GRCh38
rs78642785 Chr.5: 81330400-81330400 on Build GRCh38

View Full Product Details