Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144383054-144383309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK5
Entrez Gene ID
203054
Gene Name
aarF domain containing kinase 5
Gene Aliases
-
UniGene
Hs.283374
Gene Chromosome Location
Chr.8: 144373762-144393242 on Build GRCh38
Transcripts
NM_174922.4 XM_006716527.2 XM_011516907.2 XM_011516909.1 XM_011516910.1 XM_017013174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5409084 c.184C>T p.L62F Chr.8: 144383148-144383148 on Build GRCh38
5409083 c.183C>T p.L61L Chr.8: 144383147-144383147 on Build GRCh38
5409082 c.141G>A p.W47* Chr.8: 144383105-144383105 on Build GRCh38
6365697 c.190G>T p.A64S Chr.8: 144383154-144383154 on Build GRCh38
6324600 c.117-9T>A p.? Chr.8: 144383072-144383072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373751246 Chr.8: 144383122-144383122 on Build GRCh38
rs782196026 Chr.8: 144383261-144383261 on Build GRCh38
rs898956169 Chr.8: 144383210-144383210 on Build GRCh38
rs970672558 Chr.8: 144383095-144383095 on Build GRCh38
rs782600324 Chr.8: 144383148-144383148 on Build GRCh38
rs782060730 Chr.8: 144383081-144383081 on Build GRCh38
rs782084759 Chr.8: 144383273-144383273 on Build GRCh38
rs146980388 Chr.8: 144383138-144383138 on Build GRCh38
rs373946038 Chr.8: 144383264-144383264 on Build GRCh38
rs376959340 Chr.8: 144383176-144383176 on Build GRCh38
rs376065428 Chr.8: 144383265-144383265 on Build GRCh38
rs200675832 Chr.8: 144383274-144383274 on Build GRCh38
rs782167136 Chr.8: 144383281-144383281 on Build GRCh38
rs782385855 Chr.8: 144383175-144383175 on Build GRCh38
rs782353064 Chr.8: 144383256-144383256 on Build GRCh38
rs782747631 Chr.8: 144383110-144383110 on Build GRCh38
rs782488454 Chr.8: 144383142-144383142 on Build GRCh38
rs1026188614 Chr.8: 144383247-144383247 on Build GRCh38
rs138040309 Chr.8: 144383150-144383150 on Build GRCh38
rs989266141 Chr.8: 144383131-144383131 on Build GRCh38
rs571879889 Chr.8: 144383144-144383144 on Build GRCh38
rs781787530 Chr.8: 144383125-144383125 on Build GRCh38
rs782709627 Chr.8: 144383225-144383226 on Build GRCh38
rs782744529 Chr.8: 144383179-144383179 on Build GRCh38
rs782127102 Chr.8: 144383108-144383108 on Build GRCh38
rs914999119 Chr.8: 144383285-144383285 on Build GRCh38
rs141085129 Chr.8: 144383197-144383197 on Build GRCh38
rs781830873 Chr.8: 144383213-144383213 on Build GRCh38
rs782601775 Chr.8: 144383257-144383257 on Build GRCh38
rs375623827 Chr.8: 144383252-144383252 on Build GRCh38
rs782012182 Chr.8: 144383097-144383097 on Build GRCh38
rs782807623 Chr.8: 144383073-144383073 on Build GRCh38
rs782047392 Chr.8: 144383196-144383196 on Build GRCh38
rs781975423 Chr.8: 144383270-144383270 on Build GRCh38
rs542022535 Chr.8: 144383137-144383137 on Build GRCh38
rs1042321502 Chr.8: 144383145-144383145 on Build GRCh38
rs201452935 Chr.8: 144383245-144383245 on Build GRCh38
rs781906134 Chr.8: 144383243-144383243 on Build GRCh38
rs782038841 Chr.8: 144383158-144383158 on Build GRCh38
rs369954772 Chr.8: 144383178-144383178 on Build GRCh38
rs782541219 Chr.8: 144383126-144383126 on Build GRCh38
rs782782811 Chr.8: 144383157-144383157 on Build GRCh38
rs370217576 Chr.8: 144383262-144383262 on Build GRCh38
rs781844552 Chr.8: 144383248-144383249 on Build GRCh38
rs201592392 Chr.8: 144383240-144383240 on Build GRCh38
rs144883115 Chr.8: 144383199-144383199 on Build GRCh38
rs933607862 Chr.8: 144383161-144383161 on Build GRCh38
rs539604921 Chr.8: 144383275-144383275 on Build GRCh38
rs373468984 Chr.8: 144383246-144383246 on Build GRCh38
rs781932612 Chr.8: 144383191-144383191 on Build GRCh38
rs782274795 Chr.8: 144383168-144383168 on Build GRCh38

View Full Product Details