Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47590415-47590678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF4
Entrez Gene ID
114900
Gene Name
C1q and tumor necrosis factor related protein 4
Gene Aliases
CTRP4, ZACRP4
UniGene
Hs.662633
Gene Chromosome Location
Chr.11: 47589664-47596336 on Build GRCh38
Transcripts
NM_031909.2 XM_017017165.1 XM_017017166.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4889303 c.187G>A p.V63M Chr.11: 47590624-47590624 on Build GRCh38
4432685 c.298G>A p.E100K Chr.11: 47590513-47590513 on Build GRCh38
1981866 c.150C>T p.F50F Chr.11: 47590661-47590661 on Build GRCh38
1981863 c.201G>A p.Q67Q Chr.11: 47590610-47590610 on Build GRCh38
5951893 c.348C>T p.R116R Chr.11: 47590463-47590463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748981808 Chr.11: 47590552-47590553 on Build GRCh38
rs775181056 Chr.11: 47590616-47590616 on Build GRCh38
rs200741469 Chr.11: 47590562-47590562 on Build GRCh38
rs757153632 Chr.11: 47590542-47590542 on Build GRCh38
rs776876800 Chr.11: 47590443-47590443 on Build GRCh38
rs912016036 Chr.11: 47590448-47590448 on Build GRCh38
rs138363785 Chr.11: 47590637-47590637 on Build GRCh38
rs1042988606 Chr.11: 47590520-47590520 on Build GRCh38
rs759932330 Chr.11: 47590595-47590595 on Build GRCh38
rs769772631 Chr.11: 47590577-47590577 on Build GRCh38
rs773645943 Chr.11: 47590481-47590481 on Build GRCh38
rs535287780 Chr.11: 47590653-47590653 on Build GRCh38
rs763517517 Chr.11: 47590623-47590623 on Build GRCh38
rs11537581 Chr.11: 47590648-47590648 on Build GRCh38
rs945920244 Chr.11: 47590503-47590503 on Build GRCh38
rs779933120 Chr.11: 47590642-47590642 on Build GRCh38
rs755687320 Chr.11: 47590513-47590513 on Build GRCh38
rs867047470 Chr.11: 47590569-47590569 on Build GRCh38
rs189005009 Chr.11: 47590502-47590502 on Build GRCh38
rs773272876 Chr.11: 47590579-47590579 on Build GRCh38
rs1052460403 Chr.11: 47590469-47590469 on Build GRCh38
rs778606099 Chr.11: 47590555-47590555 on Build GRCh38
rs765271573 Chr.11: 47590631-47590631 on Build GRCh38
rs748381810 Chr.11: 47590575-47590575 on Build GRCh38
rs367790771 Chr.11: 47590598-47590598 on Build GRCh38
rs745611624 Chr.11: 47590561-47590561 on Build GRCh38
rs750358585 Chr.11: 47590634-47590634 on Build GRCh38
rs763402520 Chr.11: 47590466-47590466 on Build GRCh38
rs867417959 Chr.11: 47590610-47590610 on Build GRCh38
rs753350563 Chr.11: 47590625-47590625 on Build GRCh38
rs749788591 Chr.11: 47590580-47590580 on Build GRCh38
rs761680898 Chr.11: 47590629-47590629 on Build GRCh38
rs767863057 Chr.11: 47590601-47590601 on Build GRCh38
rs763455674 Chr.11: 47590482-47590482 on Build GRCh38
rs944677492 Chr.11: 47590454-47590454 on Build GRCh38
rs748291853 Chr.11: 47590473-47590473 on Build GRCh38
rs767205996 Chr.11: 47590496-47590496 on Build GRCh38
rs868389992 Chr.11: 47590525-47590525 on Build GRCh38
rs769890642 Chr.11: 47590475-47590475 on Build GRCh38
rs913922505 Chr.11: 47590484-47590484 on Build GRCh38
rs904318115 Chr.11: 47590563-47590563 on Build GRCh38
rs763647438 Chr.11: 47590518-47590518 on Build GRCh38
rs781609630 Chr.11: 47590568-47590568 on Build GRCh38
rs753733634 Chr.11: 47590540-47590540 on Build GRCh38
rs576074211 Chr.11: 47590456-47590456 on Build GRCh38
rs774735740 Chr.11: 47590492-47590492 on Build GRCh38
rs768962525 Chr.11: 47590438-47590438 on Build GRCh38
rs766654827 Chr.11: 47590490-47590490 on Build GRCh38
rs759206742 Chr.11: 47590493-47590493 on Build GRCh38
rs768261291 Chr.11: 47590576-47590578 on Build GRCh38
rs760318749 Chr.11: 47590621-47590621 on Build GRCh38
rs774815192 Chr.11: 47590591-47590591 on Build GRCh38
rs771417491 Chr.11: 47590581-47590581 on Build GRCh38

View Full Product Details