Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13762896-13763145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC130
Entrez Gene ID
81576
Gene Name
coiled-coil domain containing 130
Gene Aliases
-
UniGene
Hs.24998
Gene Chromosome Location
Chr.19: 13731760-13763298 on Build GRCh38
Transcripts
NM_001320561.1 NM_001320564.1 NM_001320565.1 NM_001320566.1 NM_001320567.1 NM_001320568.1 NM_001320569.1 NM_030818.3 XM_005260086.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6374460 c.1170C>A p.Y390* Chr.19: 13763047-13763047 on Build GRCh38
7448487 c.1067G>A p.R356H Chr.19: 13762944-13762944 on Build GRCh38
4909902 c.1179G>A p.S393S Chr.19: 13763056-13763056 on Build GRCh38
6386489 c.1076A>T p.K359M Chr.19: 13762953-13762953 on Build GRCh38
4459288 c.1119C>T p.T373T Chr.19: 13762996-13762996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773075861 Chr.19: 13762948-13762948 on Build GRCh38
rs757671150 Chr.19: 13762968-13762968 on Build GRCh38
rs35254119 Chr.19: 13763056-13763056 on Build GRCh38
rs776699025 Chr.19: 13763000-13763000 on Build GRCh38
rs754578587 Chr.19: 13762975-13762975 on Build GRCh38
rs758138254 Chr.19: 13762994-13762994 on Build GRCh38
rs771859681 Chr.19: 13762944-13762944 on Build GRCh38
rs763356217 Chr.19: 13762962-13762962 on Build GRCh38
rs757972465 Chr.19: 13763084-13763084 on Build GRCh38
rs986548882 Chr.19: 13763041-13763041 on Build GRCh38
rs925893143 Chr.19: 13763005-13763005 on Build GRCh38
rs992588151 Chr.19: 13762943-13762943 on Build GRCh38
rs752129226 Chr.19: 13762924-13762924 on Build GRCh38
rs553243071 Chr.19: 13762974-13762974 on Build GRCh38
rs767937440 Chr.19: 13763069-13763069 on Build GRCh38
rs770762598 Chr.19: 13762999-13762999 on Build GRCh38
rs761318210 Chr.19: 13763046-13763051 on Build GRCh38
rs375166277 Chr.19: 13763096-13763096 on Build GRCh38
rs368367653 Chr.19: 13763113-13763113 on Build GRCh38
rs775176989 Chr.19: 13763111-13763111 on Build GRCh38
rs749425586 Chr.19: 13763055-13763055 on Build GRCh38
rs391318 Chr.19: 13763077-13763077 on Build GRCh38
rs1005188874 Chr.19: 13763125-13763125 on Build GRCh38
rs776953527 Chr.19: 13762993-13762998 on Build GRCh38
rs774554508 Chr.19: 13763013-13763013 on Build GRCh38
rs747207901 Chr.19: 13762958-13762959 on Build GRCh38
rs767903135 Chr.19: 13763016-13763016 on Build GRCh38
rs750940238 Chr.19: 13763017-13763017 on Build GRCh38
rs775683553 Chr.19: 13763053-13763053 on Build GRCh38
rs752075850 Chr.19: 13762964-13762964 on Build GRCh38
rs755604097 Chr.19: 13762926-13762926 on Build GRCh38
rs760106107 Chr.19: 13762997-13763005 on Build GRCh38
rs752977270 Chr.19: 13762960-13762960 on Build GRCh38
rs764748054 Chr.19: 13763022-13763022 on Build GRCh38
rs771253180 Chr.19: 13762980-13762982 on Build GRCh38
rs767015262 Chr.19: 13763049-13763050 on Build GRCh38
rs760351746 Chr.19: 13763093-13763093 on Build GRCh38
rs781075524 Chr.19: 13763107-13763107 on Build GRCh38
rs775919335 Chr.19: 13763030-13763051 on Build GRCh38
rs773543515 Chr.19: 13763076-13763076 on Build GRCh38
rs761199578 Chr.19: 13763080-13763080 on Build GRCh38
rs747810462 Chr.19: 13762986-13762986 on Build GRCh38
rs746872128 Chr.19: 13762997-13762997 on Build GRCh38
rs766910046 Chr.19: 13763082-13763082 on Build GRCh38
rs113411146 Chr.19: 13763122-13763122 on Build GRCh38
rs770273528 Chr.19: 13763029-13763046 on Build GRCh38
rs781745414 Chr.19: 13762971-13762971 on Build GRCh38
rs780937753 Chr.19: 13763049-13763049 on Build GRCh38
rs774697198 Chr.19: 13763059-13763059 on Build GRCh38
rs201523259 Chr.19: 13762963-13762963 on Build GRCh38
rs762013876 Chr.19: 13763062-13763062 on Build GRCh38
rs757080143 Chr.19: 13763042-13763042 on Build GRCh38
rs748867066 Chr.19: 13762930-13762930 on Build GRCh38
rs149179548 Chr.19: 13763004-13763004 on Build GRCh38
rs557057122 Chr.19: 13763029-13763029 on Build GRCh38
rs756956333 Chr.19: 13763095-13763095 on Build GRCh38
rs758013394 Chr.19: 13763023-13763023 on Build GRCh38
rs760550949 Chr.19: 13762954-13762954 on Build GRCh38
rs776553036 Chr.19: 13762956-13762956 on Build GRCh38
rs747929593 Chr.19: 13762938-13762938 on Build GRCh38
rs777859304 Chr.19: 13762925-13762926 on Build GRCh38
rs778361897 Chr.19: 13762922-13762922 on Build GRCh38
rs769957451 Chr.19: 13763051-13763051 on Build GRCh38
rs763301727 Chr.19: 13763011-13763011 on Build GRCh38
rs750226979 Chr.19: 13763108-13763108 on Build GRCh38
rs754427210 Chr.19: 13763020-13763020 on Build GRCh38
rs752191958 Chr.19: 13763083-13763083 on Build GRCh38
rs778591283 Chr.19: 13762937-13762937 on Build GRCh38
rs772760651 Chr.19: 13763059-13763062 on Build GRCh38
rs756101319 Chr.19: 13763109-13763109 on Build GRCh38
rs755321670 Chr.19: 13763014-13763014 on Build GRCh38
rs771000575 Chr.19: 13762955-13762955 on Build GRCh38
rs759720626 Chr.19: 13762959-13762959 on Build GRCh38
rs751239001 Chr.19: 13763091-13763091 on Build GRCh38
rs957377883 Chr.19: 13763032-13763032 on Build GRCh38
rs138278370 Chr.19: 13762979-13762979 on Build GRCh38
rs372266888 Chr.19: 13763007-13763007 on Build GRCh38
rs779463722 Chr.19: 13762927-13762927 on Build GRCh38
rs746777103 Chr.19: 13763033-13763033 on Build GRCh38
rs763587761 Chr.19: 13763089-13763089 on Build GRCh38

View Full Product Details