Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 247111413-247111644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf229
Entrez Gene ID
388759
Gene Name
chromosome 1 open reading frame 229
Gene Aliases
-
UniGene
Hs.456511
Gene Chromosome Location
Chr.1: 247110160-247112417 on Build GRCh38
Transcripts
NM_207401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4143628 c.628T>C p.S210P Chr.1: 247111597-247111597 on Build GRCh38
4667379 c.696G>A p.P232P Chr.1: 247111529-247111529 on Build GRCh38
5761665 c.681C>T p.A227A Chr.1: 247111544-247111544 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765211437 Chr.1: 247111583-247111583 on Build GRCh38
rs938291687 Chr.1: 247111609-247111609 on Build GRCh38
rs865950230 Chr.1: 247111540-247111540 on Build GRCh38
rs545987988 Chr.1: 247111552-247111552 on Build GRCh38
rs554036898 Chr.1: 247111468-247111468 on Build GRCh38
rs921113031 Chr.1: 247111439-247111439 on Build GRCh38
rs946077976 Chr.1: 247111527-247111527 on Build GRCh38
rs562909507 Chr.1: 247111559-247111559 on Build GRCh38
rs762983239 Chr.1: 247111542-247111542 on Build GRCh38
rs540573120 Chr.1: 247111451-247111451 on Build GRCh38
rs973845001 Chr.1: 247111507-247111507 on Build GRCh38
rs763879093 Chr.1: 247111558-247111558 on Build GRCh38
rs975102235 Chr.1: 247111436-247111436 on Build GRCh38
rs769166861 Chr.1: 247111524-247111524 on Build GRCh38
rs779569228 Chr.1: 247111614-247111614 on Build GRCh38
rs966839314 Chr.1: 247111572-247111572 on Build GRCh38
rs143099468 Chr.1: 247111537-247111537 on Build GRCh38
rs954762169 Chr.1: 247111432-247111432 on Build GRCh38
rs908382955 Chr.1: 247111574-247111574 on Build GRCh38
rs868065159 Chr.1: 247111564-247111564 on Build GRCh38
rs746633022 Chr.1: 247111520-247111520 on Build GRCh38
rs369865565 Chr.1: 247111581-247111581 on Build GRCh38
rs777079527 Chr.1: 247111525-247111525 on Build GRCh38
rs957855551 Chr.1: 247111462-247111462 on Build GRCh38
rs762261904 Chr.1: 247111529-247111529 on Build GRCh38
rs913810878 Chr.1: 247111514-247111514 on Build GRCh38
rs773833205 Chr.1: 247111538-247111538 on Build GRCh38
rs750348637 Chr.1: 247111593-247111593 on Build GRCh38
rs779612949 Chr.1: 247111465-247111465 on Build GRCh38
rs746656444 Chr.1: 247111623-247111623 on Build GRCh38
rs73135916 Chr.1: 247111597-247111597 on Build GRCh38
rs868474806 Chr.1: 247111543-247111543 on Build GRCh38
rs759742714 Chr.1: 247111555-247111555 on Build GRCh38
rs865993879 Chr.1: 247111545-247111545 on Build GRCh38
rs571928030 Chr.1: 247111438-247111438 on Build GRCh38
rs760920379 Chr.1: 247111548-247111549 on Build GRCh38
rs964737741 Chr.1: 247111489-247111489 on Build GRCh38
rs766438591 Chr.1: 247111548-247111548 on Build GRCh38
rs753702586 Chr.1: 247111579-247111579 on Build GRCh38
rs531848129 Chr.1: 247111571-247111571 on Build GRCh38
rs1019343990 Chr.1: 247111429-247111429 on Build GRCh38
rs754741653 Chr.1: 247111627-247111627 on Build GRCh38
rs867899626 Chr.1: 247111551-247111551 on Build GRCh38
rs887351942 Chr.1: 247111604-247111604 on Build GRCh38
rs982588314 Chr.1: 247111553-247111553 on Build GRCh38
rs868800360 Chr.1: 247111539-247111539 on Build GRCh38

View Full Product Details