Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49219809-49220034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI3
Entrez Gene ID
51225
Gene Name
ABI family member 3
Gene Aliases
NESH, SSH3BP3
UniGene
Hs.130719
Gene Chromosome Location
Chr.17: 49210227-49223225 on Build GRCh38
Transcripts
NM_001135186.1 NM_016428.2 XM_005257429.2 XM_011524873.1 XM_017024721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5732484 c.560G>A p.R187Q Chr.17: 49219869-49219869 on Build GRCh38
4769592 c.613G>A p.A205T Chr.17: 49219922-49219922 on Build GRCh38
6590975 c.623C>T p.A208V Chr.17: 49219932-49219932 on Build GRCh38
4130358 c.626T>C p.F209S Chr.17: 49219935-49219935 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746593958 Chr.17: 49219926-49219926 on Build GRCh38
rs573287028 Chr.17: 49219908-49219908 on Build GRCh38
rs755161200 Chr.17: 49219894-49219894 on Build GRCh38
rs759026836 Chr.17: 49219848-49219848 on Build GRCh38
rs768409072 Chr.17: 49219931-49219931 on Build GRCh38
rs762812946 Chr.17: 49219943-49219943 on Build GRCh38
rs751587712 Chr.17: 49219952-49219952 on Build GRCh38
rs960287408 Chr.17: 49220011-49220011 on Build GRCh38
rs753678829 Chr.17: 49219864-49219864 on Build GRCh38
rs774865080 Chr.17: 49219921-49219921 on Build GRCh38
rs768276754 Chr.17: 49219868-49219869 on Build GRCh38
rs773638825 Chr.17: 49219871-49219874 on Build GRCh38
rs879250804 Chr.17: 49219835-49219835 on Build GRCh38
rs780020209 Chr.17: 49219889-49219889 on Build GRCh38
rs1048385507 Chr.17: 49219911-49219911 on Build GRCh38
rs761757074 Chr.17: 49219866-49219866 on Build GRCh38
rs112224699 Chr.17: 49220006-49220006 on Build GRCh38
rs760086794 Chr.17: 49219846-49219847 on Build GRCh38
rs771548372 Chr.17: 49219919-49219919 on Build GRCh38
rs766418050 Chr.17: 49219947-49219947 on Build GRCh38
rs763877827 Chr.17: 49219860-49219860 on Build GRCh38
rs773771344 Chr.17: 49219847-49219847 on Build GRCh38
rs199635084 Chr.17: 49219932-49219932 on Build GRCh38
rs760256482 Chr.17: 49219853-49219853 on Build GRCh38
rs752960723 Chr.17: 49219981-49219981 on Build GRCh38
rs1016412324 Chr.17: 49219920-49219920 on Build GRCh38
rs771525248 Chr.17: 49219849-49219849 on Build GRCh38
rs750425949 Chr.17: 49219879-49219879 on Build GRCh38
rs748501649 Chr.17: 49219907-49219907 on Build GRCh38
rs770284225 Chr.17: 49219836-49219836 on Build GRCh38
rs765243659 Chr.17: 49219877-49219877 on Build GRCh38
rs616338 Chr.17: 49219935-49219935 on Build GRCh38
rs775156826 Chr.17: 49219850-49219850 on Build GRCh38
rs767707040 Chr.17: 49219972-49219972 on Build GRCh38
rs749794758 Chr.17: 49219918-49219918 on Build GRCh38
rs868437723 Chr.17: 49219922-49219922 on Build GRCh38
rs781461563 Chr.17: 49219906-49219906 on Build GRCh38
rs756433989 Chr.17: 49219995-49219995 on Build GRCh38
rs775253151 Chr.17: 49219847-49219847 on Build GRCh38
rs751746635 Chr.17: 49219890-49219890 on Build GRCh38
rs868573721 Chr.17: 49219940-49219940 on Build GRCh38
rs367805289 Chr.17: 49219987-49219987 on Build GRCh38
rs906347959 Chr.17: 49219986-49219986 on Build GRCh38
rs765153814 Chr.17: 49219933-49219933 on Build GRCh38
rs758463151 Chr.17: 49219887-49219887 on Build GRCh38
rs755147514 Chr.17: 49219959-49219959 on Build GRCh38

View Full Product Details