Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73492703-73492974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf169
Entrez Gene ID
79697
Gene Name
chromosome 14 open reading frame 169
Gene Aliases
MAPJD, NO66, ROX, URLC2, hsNO66
UniGene
Hs.509916
Gene Chromosome Location
Chr.14: 73490934-73493401 on Build GRCh38
Transcripts
NM_024644.4 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

Gene Symbol
HEATR4
Entrez Gene ID
399671
Gene Name
HEAT repeat containing 4
Gene Aliases
-
UniGene
Hs.660774
Gene Chromosome Location
Chr.14: 73478484-73634418 on Build GRCh38
Transcripts
NM_024644.4 NM_001220484.1 NM_203309.2 XM_006720143.2 XM_011536760.2 XM_017021289.1

SNP Detail

SNP ID Chromosome Location
rs375389059 Chr.14: 73492887-73492887 on Build GRCh38
rs966105250 Chr.14: 73492941-73492941 on Build GRCh38
rs1032189656 Chr.14: 73492878-73492878 on Build GRCh38
rs779248928 Chr.14: 73492751-73492751 on Build GRCh38
rs775347915 Chr.14: 73492935-73492936 on Build GRCh38
rs754013856 Chr.14: 73492848-73492848 on Build GRCh38
rs760477509 Chr.14: 73492823-73492823 on Build GRCh38
rs200061659 Chr.14: 73492870-73492870 on Build GRCh38
rs151064235 Chr.14: 73492861-73492861 on Build GRCh38
rs781647964 Chr.14: 73492939-73492939 on Build GRCh38
rs367909112 Chr.14: 73492784-73492784 on Build GRCh38
rs3742846 Chr.14: 73492892-73492892 on Build GRCh38
rs750383577 Chr.14: 73492815-73492815 on Build GRCh38
rs754132267 Chr.14: 73492737-73492737 on Build GRCh38
rs747408546 Chr.14: 73492779-73492779 on Build GRCh38
rs771304403 Chr.14: 73492925-73492925 on Build GRCh38
rs576690817 Chr.14: 73492730-73492730 on Build GRCh38
rs768345086 Chr.14: 73492800-73492800 on Build GRCh38
rs755214593 Chr.14: 73492746-73492746 on Build GRCh38
rs905113343 Chr.14: 73492850-73492850 on Build GRCh38
rs373901476 Chr.14: 73492808-73492808 on Build GRCh38
rs748855069 Chr.14: 73492951-73492951 on Build GRCh38
rs771987519 Chr.14: 73492839-73492862 on Build GRCh38
rs755089726 Chr.14: 73492857-73492857 on Build GRCh38
rs766508250 Chr.14: 73492841-73492841 on Build GRCh38
rs199717657 Chr.14: 73492802-73492802 on Build GRCh38
rs762418034 Chr.14: 73492790-73492790 on Build GRCh38
rs772314263 Chr.14: 73492761-73492761 on Build GRCh38
rs200600929 Chr.14: 73492924-73492924 on Build GRCh38
rs776661834 Chr.14: 73492916-73492916 on Build GRCh38
rs769362135 Chr.14: 73492780-73492780 on Build GRCh38
rs778412349 Chr.14: 73492762-73492762 on Build GRCh38
rs1020774924 Chr.14: 73492932-73492932 on Build GRCh38
rs34568526 Chr.14: 73492816-73492817 on Build GRCh38
rs748576282 Chr.14: 73492752-73492752 on Build GRCh38

View Full Product Details