Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56370432-56370689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKA
Entrez Gene ID
6790
Gene Name
aurora kinase A
Gene Aliases
AIK, ARK1, AURA, BTAK, PPP1R47, STK15, STK6, STK7
UniGene
Hs.250822
Gene Chromosome Location
Chr.20: 56369389-56392337 on Build GRCh38
Transcripts
NM_001323303.1 NM_001323304.1 NM_001323305.1 NM_003600.3 NM_198433.2 NM_198434.2 NM_198435.2 NM_198436.2 NM_198437.2 XM_017028034.1 XM_017028035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6864136 c.882C>A p.D294E Chr.20: 56370632-56370632 on Build GRCh38
6902659 c.860C>T p.T287I Chr.20: 56370654-56370654 on Build GRCh38
7268579 c.1021A>G p.I341V Chr.20: 56370493-56370493 on Build GRCh38
1307535 c.969A>C p.L323F Chr.20: 56370545-56370545 on Build GRCh38
6608851 c.910C>T p.R304W Chr.20: 56370604-56370604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778060524 Chr.20: 56370477-56370477 on Build GRCh38
rs978663717 Chr.20: 56370555-56370555 on Build GRCh38
rs11539196 Chr.20: 56370541-56370541 on Build GRCh38
rs779238744 Chr.20: 56370634-56370634 on Build GRCh38
rs772720875 Chr.20: 56370654-56370654 on Build GRCh38
rs1031527931 Chr.20: 56370596-56370596 on Build GRCh38
rs774537899 Chr.20: 56370646-56370647 on Build GRCh38
rs774552788 Chr.20: 56370487-56370487 on Build GRCh38
rs772280719 Chr.20: 56370489-56370489 on Build GRCh38
rs759640621 Chr.20: 56370488-56370488 on Build GRCh38
rs928987283 Chr.20: 56370535-56370535 on Build GRCh38
rs750222400 Chr.20: 56370524-56370524 on Build GRCh38
rs560948705 Chr.20: 56370642-56370642 on Build GRCh38
rs200173136 Chr.20: 56370494-56370494 on Build GRCh38
rs373216908 Chr.20: 56370506-56370506 on Build GRCh38
rs761713577 Chr.20: 56370507-56370507 on Build GRCh38
rs772264070 Chr.20: 56370668-56370668 on Build GRCh38
rs762567231 Chr.20: 56370550-56370550 on Build GRCh38
rs986197160 Chr.20: 56370667-56370667 on Build GRCh38
rs200181472 Chr.20: 56370486-56370486 on Build GRCh38
rs751240542 Chr.20: 56370593-56370593 on Build GRCh38
rs762209041 Chr.20: 56370668-56370670 on Build GRCh38
rs776571850 Chr.20: 56370500-56370500 on Build GRCh38
rs756649666 Chr.20: 56370604-56370604 on Build GRCh38
rs922420843 Chr.20: 56370617-56370617 on Build GRCh38
rs34728205 Chr.20: 56370505-56370506 on Build GRCh38
rs774112994 Chr.20: 56370518-56370518 on Build GRCh38
rs753238269 Chr.20: 56370603-56370603 on Build GRCh38
rs775499369 Chr.20: 56370669-56370669 on Build GRCh38
rs765075405 Chr.20: 56370516-56370516 on Build GRCh38
rs371787863 Chr.20: 56370612-56370612 on Build GRCh38
rs770142049 Chr.20: 56370473-56370473 on Build GRCh38
rs748610305 Chr.20: 56370462-56370462 on Build GRCh38
rs200774436 Chr.20: 56370478-56370478 on Build GRCh38
rs1046125726 Chr.20: 56370546-56370546 on Build GRCh38
rs771693741 Chr.20: 56370626-56370626 on Build GRCh38
rs765964564 Chr.20: 56370592-56370592 on Build GRCh38
rs780584099 Chr.20: 56370599-56370599 on Build GRCh38

View Full Product Details