Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 178521107-178521380 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX35
Entrez Gene ID
84066
Gene Name
testis expressed 35
Gene Aliases
C1orf49, TSC24
UniGene
Hs.534501
Gene Chromosome Location
Chr.1: 178513077-178523500 on Build GRCh38
Transcripts
NM_001170722.1 NM_001170723.1 NM_001170724.1 NM_032126.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1133890 c.572A>G p.N191S Chr.1: 178521250-178521250 on Build GRCh38
6800303 c.584G>A p.R195Q Chr.1: 178521262-178521262 on Build GRCh38
3418402 c.49G>A p.E17K Chr.1: 178521224-178521224 on Build GRCh38
1473054 c.583C>T p.R195W Chr.1: 178521261-178521261 on Build GRCh38
5002806 c.342G>T p.K114N Chr.1: 178521248-178521248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775081632 Chr.1: 178521196-178521196 on Build GRCh38
rs981996535 Chr.1: 178521324-178521324 on Build GRCh38
rs767978232 Chr.1: 178521291-178521291 on Build GRCh38
rs763854246 Chr.1: 178521205-178521205 on Build GRCh38
rs1002072498 Chr.1: 178521345-178521345 on Build GRCh38
rs567443882 Chr.1: 178521152-178521152 on Build GRCh38
rs891795036 Chr.1: 178521321-178521321 on Build GRCh38
rs371900017 Chr.1: 178521278-178521278 on Build GRCh38
rs746543349 Chr.1: 178521298-178521298 on Build GRCh38
rs79662719 Chr.1: 178521340-178521340 on Build GRCh38
rs200527779 Chr.1: 178521306-178521306 on Build GRCh38
rs574378762 Chr.1: 178521322-178521322 on Build GRCh38
rs906372152 Chr.1: 178521343-178521343 on Build GRCh38
rs750556920 Chr.1: 178521255-178521255 on Build GRCh38
rs900801507 Chr.1: 178521186-178521186 on Build GRCh38
rs187831526 Chr.1: 178521281-178521281 on Build GRCh38
rs201449108 Chr.1: 178521264-178521264 on Build GRCh38
rs368087165 Chr.1: 178521339-178521339 on Build GRCh38
rs879175243 Chr.1: 178521232-178521232 on Build GRCh38
rs547621564 Chr.1: 178521137-178521137 on Build GRCh38
rs954701860 Chr.1: 178521247-178521247 on Build GRCh38
rs367935438 Chr.1: 178521304-178521304 on Build GRCh38
rs765242840 Chr.1: 178521188-178521188 on Build GRCh38
rs192201686 Chr.1: 178521262-178521262 on Build GRCh38
rs1006376007 Chr.1: 178521163-178521163 on Build GRCh38
rs779315286 Chr.1: 178521274-178521274 on Build GRCh38
rs201230647 Chr.1: 178521210-178521210 on Build GRCh38
rs146319926 Chr.1: 178521259-178521259 on Build GRCh38
rs759445759 Chr.1: 178521179-178521179 on Build GRCh38
rs533799319 Chr.1: 178521265-178521265 on Build GRCh38
rs202003188 Chr.1: 178521174-178521174 on Build GRCh38
rs757596679 Chr.1: 178521224-178521224 on Build GRCh38
rs780611383 Chr.1: 178521261-178521261 on Build GRCh38
rs184090793 Chr.1: 178521280-178521280 on Build GRCh38
rs926257046 Chr.1: 178521283-178521283 on Build GRCh38
rs762820965 Chr.1: 178521204-178521204 on Build GRCh38
rs996343309 Chr.1: 178521193-178521193 on Build GRCh38
rs777206782 Chr.1: 178521282-178521282 on Build GRCh38
rs909668637 Chr.1: 178521355-178521355 on Build GRCh38
rs35619308 Chr.1: 178521252-178521252 on Build GRCh38
rs371807679 Chr.1: 178521307-178521307 on Build GRCh38
rs200548576 Chr.1: 178521297-178521297 on Build GRCh38
rs969301975 Chr.1: 178521319-178521319 on Build GRCh38

View Full Product Details