Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 26765990-26766251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRA1A
Entrez Gene ID
148
Gene Name
adrenoceptor alpha 1A
Gene Aliases
ADRA1C, ADRA1L1, ALPHA1AAR
UniGene
Hs.52931
Gene Chromosome Location
Chr.8: 26738113-26870994 on Build GRCh38
Transcripts
NM_001322502.1 NM_001322503.1 NM_033302.3 NM_033303.4 NM_033304.3 NR_136343.1 XM_006716292.3 XM_006716293.3 XM_017013094.1 XM_017013095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6991048 c.1281T>C p.D427D Chr.8: 26766138-26766138 on Build GRCh38
144096 c.1271G>A p.R424K Chr.8: 26766148-26766148 on Build GRCh38
454362 c.1338G>T p.M446I Chr.8: 26766081-26766081 on Build GRCh38
1699871 c.1307G>A p.R436K Chr.8: 26766112-26766112 on Build GRCh38
333489 c.1340C>T p.P447L Chr.8: 26766079-26766079 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757382695 Chr.8: 26766187-26766187 on Build GRCh38
rs753179722 Chr.8: 26766167-26766167 on Build GRCh38
rs61759725 Chr.8: 26766136-26766136 on Build GRCh38
rs1011806638 Chr.8: 26766039-26766039 on Build GRCh38
rs558657608 Chr.8: 26766203-26766203 on Build GRCh38
rs776488513 Chr.8: 26766102-26766102 on Build GRCh38
rs35838179 Chr.8: 26766181-26766182 on Build GRCh38
rs568851924 Chr.8: 26766209-26766209 on Build GRCh38
rs753930956 Chr.8: 26766181-26766181 on Build GRCh38
rs759757560 Chr.8: 26766161-26766161 on Build GRCh38
rs562736222 Chr.8: 26766016-26766016 on Build GRCh38
rs746107545 Chr.8: 26766190-26766190 on Build GRCh38
rs767929088 Chr.8: 26766166-26766166 on Build GRCh38
rs778472185 Chr.8: 26766172-26766172 on Build GRCh38
rs755397079 Chr.8: 26766032-26766032 on Build GRCh38
rs199526417 Chr.8: 26766170-26766170 on Build GRCh38
rs61759726 Chr.8: 26766156-26766156 on Build GRCh38
rs779228979 Chr.8: 26766093-26766093 on Build GRCh38
rs960306452 Chr.8: 26766110-26766110 on Build GRCh38
rs142383582 Chr.8: 26766076-26766076 on Build GRCh38
rs762584564 Chr.8: 26766024-26766024 on Build GRCh38
rs746068869 Chr.8: 26766123-26766123 on Build GRCh38
rs780955012 Chr.8: 26766191-26766191 on Build GRCh38
rs985732369 Chr.8: 26766215-26766215 on Build GRCh38
rs571036425 Chr.8: 26766189-26766189 on Build GRCh38
rs771285520 Chr.8: 26766181-26766184 on Build GRCh38
rs1014641207 Chr.8: 26766149-26766149 on Build GRCh38
rs781498946 Chr.8: 26766043-26766043 on Build GRCh38
rs59765666 Chr.8: 26766124-26766124 on Build GRCh38
rs548457901 Chr.8: 26766099-26766099 on Build GRCh38
rs761739096 Chr.8: 26766113-26766113 on Build GRCh38
rs771028635 Chr.8: 26766080-26766080 on Build GRCh38
rs772700723 Chr.8: 26766027-26766027 on Build GRCh38
rs751836909 Chr.8: 26766031-26766031 on Build GRCh38
rs756195367 Chr.8: 26766060-26766060 on Build GRCh38
rs766101109 Chr.8: 26766030-26766030 on Build GRCh38
rs1021730901 Chr.8: 26766047-26766051 on Build GRCh38
rs778125890 Chr.8: 26766160-26766160 on Build GRCh38
rs537865966 Chr.8: 26766214-26766214 on Build GRCh38
rs531635341 Chr.8: 26766050-26766050 on Build GRCh38
rs772690737 Chr.8: 26766128-26766128 on Build GRCh38
rs780327805 Chr.8: 26766205-26766205 on Build GRCh38
rs538311100 Chr.8: 26766202-26766202 on Build GRCh38
rs143242407 Chr.8: 26766097-26766097 on Build GRCh38
rs765906061 Chr.8: 26766134-26766134 on Build GRCh38
rs148309211 Chr.8: 26766130-26766130 on Build GRCh38
rs373841042 Chr.8: 26766075-26766075 on Build GRCh38
rs761258809 Chr.8: 26766013-26766013 on Build GRCh38
rs932361901 Chr.8: 26766211-26766211 on Build GRCh38
rs747455439 Chr.8: 26766159-26766160 on Build GRCh38

View Full Product Details