Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73436104-73436353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFP
Entrez Gene ID
174
Gene Name
alpha fetoprotein
Gene Aliases
AFPD, FETA, HPAFP
UniGene
Hs.518808
Gene Chromosome Location
Chr.4: 73436219-73455785 on Build GRCh38
Transcripts
NM_001134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6399725 c.59C>A p.T20K Chr.4: 73436321-73436321 on Build GRCh38
1430917 c.68G>T p.R23I Chr.4: 73436330-73436330 on Build GRCh38
3605686 c.49G>A p.E17K Chr.4: 73436311-73436311 on Build GRCh38
734720 c.53C>G p.S18C Chr.4: 73436315-73436315 on Build GRCh38
1057409 c.64C>T p.H22Y Chr.4: 73436326-73436326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781117277 Chr.4: 73436248-73436248 on Build GRCh38
rs752206199 Chr.4: 73436145-73436145 on Build GRCh38
rs891813350 Chr.4: 73436197-73436197 on Build GRCh38
rs777164680 Chr.4: 73436212-73436212 on Build GRCh38
rs763153741 Chr.4: 73436298-73436298 on Build GRCh38
rs904572402 Chr.4: 73436147-73436147 on Build GRCh38
rs547954049 Chr.4: 73436141-73436141 on Build GRCh38
rs201668475 Chr.4: 73436289-73436289 on Build GRCh38
rs1000283338 Chr.4: 73436187-73436187 on Build GRCh38
rs191726190 Chr.4: 73436224-73436224 on Build GRCh38
rs772124754 Chr.4: 73436279-73436279 on Build GRCh38
rs150335619 Chr.4: 73436313-73436313 on Build GRCh38
rs200781949 Chr.4: 73436288-73436288 on Build GRCh38
rs779153641 Chr.4: 73436260-73436260 on Build GRCh38
rs759858480 Chr.4: 73436310-73436310 on Build GRCh38
rs369819257 Chr.4: 73436308-73436308 on Build GRCh38
rs773234279 Chr.4: 73436283-73436283 on Build GRCh38
rs866779359 Chr.4: 73436296-73436296 on Build GRCh38
rs372320842 Chr.4: 73436294-73436294 on Build GRCh38
rs540038566 Chr.4: 73436261-73436261 on Build GRCh38
rs372836099 Chr.4: 73436258-73436258 on Build GRCh38
rs745705495 Chr.4: 73436256-73436256 on Build GRCh38
rs751445259 Chr.4: 73436222-73436222 on Build GRCh38
rs868639778 Chr.4: 73436311-73436311 on Build GRCh38
rs545418914 Chr.4: 73436199-73436199 on Build GRCh38
rs776953401 Chr.4: 73436300-73436300 on Build GRCh38
rs752338948 Chr.4: 73436320-73436320 on Build GRCh38
rs1031653196 Chr.4: 73436194-73436194 on Build GRCh38
rs766549436 Chr.4: 73436322-73436330 on Build GRCh38
rs866753077 Chr.4: 73436226-73436226 on Build GRCh38

View Full Product Details